Key Documents
EP0030128990
Eppendorf® Twin tec PCR plates, 96 wells
yellow plate, conical bottom colorless wells, skirted, pkg of 300 ea
Select a Size
300 EA
€2,580.00
€2,580.00
Check Cart for Availability
Select a Size
Change View
300 EA
€2,580.00
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
conical bottom colorless wells
yellow plate
size:
96 wells
binding type:
low binding surface
feature:
skirt
€2,580.00
Check Cart for Availability
material
conical bottom colorless wells
yellow plate
feature
skirt
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128990
maximum volume
-250 μL
size
96 wells
working volume
-200 μL
suitability
suitable for PCR
suitable for molecular biology
binding type
low binding surface
General description
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
The Eppendorf “twin.tec” PCR plates are designed for single-use PCR applications. The plates are skirted and equipped with thin-walled polypropylene wells and polycarbonate frames. These features help ensure stable handling and improved capability to withstand high temperature.
Application
“Eppendorf” Twin tec PCR plates are suitable for use in:
  • Standard PCR
  • Real-time PCR
  • High Throughput PCR
  • Small Volume Sample Handling
Features and Benefits
  • Compatible for use in automated systems.
  • The PCR plates are skirted, allowing for easy labeling or barcode placement.
  • They conform to the SBS footprint recommendations.
  • The plates are stackable, optimizing storage space.
  • With a low-profile design, they are suitable for low volume PCR applications.
  • Each well has a maximum volume of 150 ?L when used with cap strips.
  • Certified to be free of human DNA, DNase, RNase, and PCR inhibitors.
  • The raised rims of the plate enhance effective sealing thereby educing the risk of leakage or contamination.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)