Key Documents
EP0030128788
Eppendorf® Twin tec PCR plates, 96 wells
yellow plate, conical bottom colorless wells, skirted, pkg of 300 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
conical bottom colorless wells
yellow plate
size:
96 wells
well maximum volume:
150 μm
feature:
skirt
material
conical bottom colorless wells
yellow plate
feature
skirt
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128788
technique(s)
PCR: suitable
size
96 wells
well maximum volume
150 μm
General description
  • Eppendorf® Twin tec PCR plates, 96 wells are single-use polycarbonate/polypropylene plates allowing trained specialists to perform PCR and cycle sequencing experiments.
  • They are used in training, routine and research laboratories in the bioscientific, industrial and chemical fields.
  • Evaporation is prevented with the aid of a variety of sealing systems.
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)