Skip to Content
Merck
All Photos(1)

Key Documents

EP0030128575

Eppendorf® Twin tec PCR plates, 96 wells

colorless plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€154.00

€154.00


Check Cart for Availability


Select a Size

Change View
25 EA
€154.00

About This Item

UNSPSC Code:
41106300
NACRES:
NB.15
material:
colorless plate
conical bottom colorless wells
polypropylene
size:
96 wells
sterility:
non-sterile
feature:
skirt

€154.00


Check Cart for Availability

material

colorless plate
conical bottom colorless wells
polypropylene

sterility

non-sterile

feature

skirt

packaging

pkg of 25 ea

manufacturer/tradename

Eppendorf® 0030128575

technique(s)

PCR: suitable

size

96 wells

well maximum volume

250 μm

Looking for similar products? Visit Product Comparison Guide

General description

Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.

Features and Benefits

  • Eppendorf® Twin tec PCR plate combine a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors.
  • Fits most thermal cyclers.
  • 250 μL max. well volume (when used with cap strips).
  • Improved well-to-well tolerance.
  • Ideal for quantitative real-time PCR as well – OptiTrack® matrix for faster sample identification and fewer pipetting errors.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service