Documents
EP0030089855
Eppendorf® Combitips Advanced
maximum volume (1 mL), yellow, Forensic DNA Grade, pkg of individually packed tips, pack of 100 ea (individually wrapped)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
material
yellow
sterility
non-sterile
feature
Forensic DNA Grade
packaging
pack of 100 ea (individually wrapped)
pkg of individually packed tips
manufacturer/tradename
Eppendorf® 0030089855
maximum volume
(1 mL)
color
yellow
Features and Benefits
  • Combitips® Advanced Tips are the ideal consumable choice for all Multipette® multi-dispenser pipettes from Eppendorf®
  • With a range of 9 volume sizes from 0.1 mL to 50 mL, they provide a wide selection of dispensing volumes, allowing increments as small as 100 nanoliters depending on the specific Combitips Advanced used.
  • The elongated Combitips Advanced in 2.5 mL, 5.0 mL, and 10 mL volumes enable easy access to the bottom of commonly used laboratory vessels.
  • Each Combitips Advanced tip is color-coded individually, allowing for quick identification of the desired tip.
  • The unique funnel shape ensures comfortable handling while also protecting gloves from damage.
  • Combitips Advanced is offered in different purity grades to support a range of applications
  • Available in different purity grades to cater to various applications, including Eppendorf Quality, PCR Clean, Forensic DNA Grade, and Biopur®.
  • ACT® labeled by the non-profit My Green Lab® to help you make an informed sustainable product choice
Legal Information
ACT is a registered trademark of Merck KGaA, Darmstadt, Germany
Biopur is a registered trademark of Eppendorf AG
Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG
My Green Lab is a registered trademark of My Green Lab, Corp. non-profit corporation