Skip to Content
Merck
All Photos(1)

Key Documents

EMU157701

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Id4

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCAGCAAATCTAGAAACGTCGTCTCGTCTTGTCATTCCAAGAGAGAGAGAGAGAGAGAGAGAGAAGGGGAAAAATAAAACTTAAATTCACTTTTACTTTTTTTGCACGTTCACGAGCATTCACCGTACGTATTCTCTTTCGTTTCTTCTTTTATGACCGCTGTGAATTGTACGTTTCTGTGGTTATTTTTATCACCCTTTTGAAGGTGCAGTTAAACTTCGAAGCTTAAGTGTTGTCGACCAGACTGCTAAGTAGAAGAGCAATCGTGAATCCAACCTTAGAGGCTACATTGTGACAAGGGAACTGTTTTTGTTTTTGAAGCTTTACTAATATACCAGAGCACTGTAGATATGTTGTTTTACATCTATTGTTTAAAATAGATGATTATAACAGGGCGGGGAACTTTTTCTCTGCAAGAATGTTACATATTGTATAGATAAGTGAGTGACATTTCATACCCTGTATATATAGAGATGTTCTATAAGTGTGAGAAAGTATATGCGCTTTAAT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Se-Jin Park et al.
Laboratory animal research, 27(4), 333-338 (2012-01-11)
The cancer stem cell (CSC) hypothesis proposes that CSCs are the root of cancer. CSC-targeted therapies may prevent cancer relapse and provide more effective treatment. The expression of aldehyde dehydrogenase 1, as assessed by the Aldefluor assay, has been recognized
Simon Junankar et al.
Nature communications, 6, 6548-6548 (2015-03-31)
Basal-like breast cancer (BLBC) is a heterogeneous disease with poor prognosis; however, its cellular origins and aetiology are poorly understood. In this study, we show that inhibitor of differentiation 4 (ID4) is a key regulator of mammary stem cell self-renewal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service