Skip to Content
Merck
All Photos(1)

Key Documents

EMU094061

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Atg7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTTTCTGCTCCTGACCTTCGCGGACCTAAAGAAGTACCACTTCTACTACTGGTTTTGCTGCCCCGCCCTCTGTCTTCCTGAGAGCATCCCTCTAATCCGGGGACCTGTGAGCTTGGATCAAAGGCTTTCACCAAAACAGATCCAGGCCCTGGAGCATGCCTATGATGATCTGTGTCGAGCCGAAGGCGTCACGGCCCTGCCCTACTTCTTATTCAAGTACGATGACGACACTGTTCTGGTCTCCTTGCTCAAACACTACAGTGATTTCTTCCAAGGTCAAAGGACAAAGATAACAGTTGGTGTGTACGATCCCTGTAACCTAGCCCAGTACCCTGGATGGCCTTTGAGGAATTTTTTGGTCCTGGCAGCCCACAGATGGAGCGGCAGTTTCCAGTCCGTTGAAGTCC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingrong Tang et al.
PloS one, 9(7), e103364-e103364 (2014-07-26)
The two major intracellular protein degradation systems, the ubiquitin-proteasome system (UPS) and autophagy, work collaboratively in many biological processes including development, apoptosis, aging, and countering oxidative injuries. We report here that, in human retinal pigment epithelial cells (RPE), ARPE-19 cells
Marco B E Schaaf et al.
Radiotherapy and oncology : journal of the European Society for Therapeutic Radiology and Oncology, 114(3), 406-412 (2015-03-18)
(Pre)clinical studies indicate that autophagy inhibition increases response to anti-cancer therapies. Although promising, due to contradicting reports, it remains unclear if radiation therapy changes autophagy activity and if autophagy inhibition changes the cellular intrinsic radiosensitivity. Discrepancies may result from different
Hong-Guang Xia et al.
The Journal of cell biology, 210(5), 705-716 (2015-09-02)
Hexokinase II (HK2), a key enzyme involved in glucose metabolism, is regulated by growth factor signaling and is required for initiation and maintenance of tumors. Here we show that metabolic stress triggered by perturbation of receptor tyrosine kinase FLT3 in
Minghua Yang et al.
PloS one, 9(5), e97242-e97242 (2014-05-14)
Autophagy is a double-edged sword in tumorigenesis and plays an important role in the resistance of cancer cells to chemotherapy. S100A8 is a member of the S100 calcium-binding protein family and plays an important role in the drug resistance of
Hongming Pan et al.
Scientific reports, 4, 6683-6683 (2014-10-21)
Autophagy is a critical survival pathway for cancer cells under conditions of stress. Thus, induction of autophagy has emerged as a drug resistance mechanism. This study is to determine whether autophagy is activated by a novel multikinase inhibitor linifanib, thereby

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service