Skip to Content
Merck
All Photos(1)

Key Documents

EMU093981

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Bmi1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TACCATGAATGGAACCAGCAACAGCCCCAGTGCTAACCACCAATCTTCCTTTGCCAGTAGACCTCGAAAATCATCACTAAATGGGTCATCAGCAACTTCATCTGGTTAGGACTGTTAAGGAAAAGATTTTTCAACCCCCTGATTTAGTTACCTTCATTCATTACAGCTTTATAGATGCTTAATACATGTGACTGTCGTCCAGTTTGCTTCCTTTTGTAGTGACTTTAAATTTGGCCATAAATGATGGACTAGATGTGATACTTCATATGGATGTTAAGTGGAAAGATTGATTCTTTCTCTAAAGAATTGGATTCTGAGAAGGATTCTGTGTTAGGAAAGATGTGAAATGATTTCTGTGACCACTGTTTGGATCTGGAAATGTTCTACAGTGGGTAGACATTGGGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Xiong et al.
Cancer biology & therapy, 16(5), 756-763 (2015-04-17)
Previous studies indicate that the role of B lymphoma Mo-MLV insertion region 1 homolog (Bmi-1) is responsible for multiple cancer progression. However, Bmi-1 in controlling gene expression in non-small cell lung cancer (NSCLC) development is not well explored. Here we
Boyang Chang et al.
Biochimica et biophysica acta, 1840(12), 3285-3291 (2014-08-26)
Bmi-1 had been found to involve in self renewal of stem cells and tumorigenesis in various malignancies. In this study, we investigated the role of Bmi-1 in the development of salivary adenoid cystic carcinoma (SACC). At first, we confirmed that
Lei Liu et al.
International journal of clinical and experimental pathology, 8(6), 6674-6682 (2015-08-12)
The aim of this study was to evaluate the efficiency of a targeted siRNA nano-delivery system to silence the expression of Bmi-1 and hTERT, and to verify the toxicity of this delivery system in MCF-7 breast cancer cells. The most
F Wei et al.
Oncogene, 34(23), 3063-3075 (2014-08-05)
The BMI1 protein contributes to stem cell pluripotency and oncogenesis via multiple functions, including its newly identified role in DNA damage response (DDR). Although evidence clearly demonstrates that BMI1 facilitates the repair of double-stranded breaks via homologous recombination (HR), it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service