Skip to Content
Merck

Skip To

EMU080301

MISSION® esiRNA

targeting mouse Cntn2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 212,00
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 382,00

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

€ 212,00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATCAGTCGATCGGATGAAGGCAAATATACCTGCTTTGCTGAGAACTTCATGGGCAAAGCCAACAGTACCGGGATCCTGTCTGTGCGAGATGCAACGAAGATCACGCTAGCTCCTTCGAGTGCTGACATCAATGTGGGTGATAACCTGACCCTACAGTGCCACGCCTCGCATGACCCCACTATGGACCTCACGTTCACCTGGACCCTGGATGACTTCCCTGTTGACTTTGATAAGCCTGGAGGTCACTACCGGAGAGCCAGTGTGAAGGAGACCATTGGGGACCTGACTATCCTGAATGCCCAGCTACGCCACGGAGGGACATACACATGCATGGCCCAGACAGTGGTGGATGGTGCATCCAAGGAGGCCACAGTCCTGGTCCGAGGTCCCCCAGGTCCTCCTGGGGGTGTGGTGGTGAGAGACATTGGAGACACTACTGTTCAGCTTAGCTGGAGTCGTGGCTTT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMU057651EMU038031EMU215231
Gene Information

mouse ... CNTN2(21367), Cntn2(21367)

Gene Information

mouse ... TBRG1(21376), Tbrg1(21376)

Gene Information

mouse ... TEX264(21767), Tex264(21767)

Gene Information

mouse ... RPL3(27367), Rpl3(27367)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

ATCAGTCGATCGGATGAAGGCAAATATACCTGCTTTGCTGAGAACTTCATGGGCAAAGCCAACAGTACCGGGATCCTGTCTGTGCGAGATGCAACGAAGATCACGCTAGCTCCTTCGAGTGCTGACATCAATGTGGGTGATAACCTGACCCTACAGTGCCACGCCTCGCATGACCCCACTATGGACCTCACGTTCACCTGGACCCTGGATGACTTCCCTGTTGACTTTGATAAGCCTGGAGGTCACTACCGGAGAGCCAGTGTGAAGGAGACCATTGGGGACCTGACTATCCTGAATGCCCAGCTACGCCACGGAGGGACATACACATGCATGGCCCAGACAGTGGTGGATGGTGCATCCAAGGAGGCCACAGTCCTGGTCCGAGGTCCCCCAGGTCCTCCTGGGGGTGTGGTGGTGAGAGACATTGGAGACACTACTGTTCAGCTTAGCTGGAGTCGTGGCTTT

esiRNA cDNA target sequence

AGTGCCCAGACCAGAAGTGTCTGTATACCTGTCAGATCAAGGACGGCGGTGTACAGCCTCAGTTTGAAATTGTTCCTGAAGATGACCCCCAGAATACCATCGTCGGCTCCTCTGCAGATGCCTGTTATGAAGAACTGCTCAGGGCCATCAGTGCCACAACAGGGAAACTGATGCCCAACCCACTTTCATGCGGTGCCGACTTCTTTGGGTTTTCTCATCCAACCATCCACAACCTGATTCAGAGTTGTCCAGAAGCTCAAAACTGTGTCAATTACCAGTGGGTGAAATTTGATGCGTGCAAACCCAGGAAGGGGCAGCTCTCTCAGGAACTGCCAGAGAATGATGCAACTATGAGCCTTGAAGCCTTCCAGACACAGACGTTTGATGATGACCATGATGATTCCATTCTGCCAGGCTCCCTGGACCTCCCTGAGCTTCAGCATGAAGCCTTTGTGTCATCTTACCAGCCAGAGTTCCTGACACACGAACCCTTGGTTGACACTGACCTGCAGCACCTGAAATCT

esiRNA cDNA target sequence

GGCACCTTTTCACAGAGAGCTGCAGCATCTCTCCCAAGCTCCGTTCCATCGCTGTCTACTATGACAACCCCCATACGGTGCCTCCTGAGAAGTGCCGCTGTGCAGTCGGCAGCATCCTGAGTGAGGNGGAGGAGTCGCCTTCACCTGAGCTCATCCACCTCTATCAGAAATTTGGCTTCAAGATATTCTCCTTCCCAGCACCTAGCCATGTGGTCATAGCTACCTTCCCTTACACCACCCCCATATCCATCTGGCTGGCTGCCCGCCGAGTCCATCCTGCCTTGGATACCTACATCAAGGAGCGGAAGCTGTGTGCTCACCCTCGCCTGGAGATCTACCATCAAGACAAGATCCATTTCATGTGCCCACTGGCAAGGCAAGGAGATTTCTACGTGCCAGAGGTGAAGGAGACAGAGCGGAAATGCCGGGAGCTTGCGGAGGCCACTGACACCCAGACGGATGGCACAGGAGCTGATACAAGTGATGCAAGTTCTGTGAGCC

esiRNA cDNA target sequence

AAAAGGCAAAGGGTACAAAGGGGTAACCAGTCGTTGGCATACAAAGAAACTGCCCCGAAAGACCCATCGAGGTTTGCGCAAAGTTGCCTGTATTGGAGCCTGGCACCCTGCCCGTGTTGCCTTCTCTGTGGCTCGAGCTGGGCAGAAAGGCTACCATCACCGAACAGAGATTAACAAGAAGATTTACAAGATTGGTCAAGGCTACCTCATCAAGGATGGCAAACTGATCAAGAACAATGCATCTACTGACTATGACTTGTCTGACAAGAGCATCAACCCACTGGGTGGCTTTGTGCATTATGGTGAGGTGACCAATGACTTCATCATGCTCAAAGGCTGTGTGGTGGGGACCAAGAAGCGAGTACTTACTCTTCGTAAGTCCTTGCTGGTTCAGACCAAACGTCGGGCCCTGGAGAAGATTGACCTGAAGTTCATTGACACCACCTCCAAATTTGGCCATGGTCGCTTCCAGACCATGGAGGAGAAGAAAGCATTTATGGGACCACTCAA

Ensembl | mouse accession no.

ENSMUSG00000053024

Ensembl | mouse accession no.

ENSMUSG00000011114

Ensembl | mouse accession no.

ENSMUSG00000040813

Ensembl | mouse accession no.

ENSMUSG00000060036

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters