Skip to Content
Merck
All Photos(1)

Documents

EMU047141

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Il1rl1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATTGCCTGTTCAGCTTGCTTTGGCAAAGGCTCTCACTTCTTGGCTGATGTCCTGTGGCAGATTAACAAAACAGTAGTTGGAAATTTTGGTGAAGCAAGAATTCAAGAAGAGGAAGGTCGAAATGAAAGTTCCAGCAATGACATGGATTGTTTAACCTCAGTGTTAAGGATAACTGGTGTGACAGAAAAGGACCTGTCCCTGGAATATGACTGTCTGGCCCTGAACCTTCATGGCATGATAAGGCACACCATAAGACTGAGAAGGAAACAACCAAGTAAGGAGTGTCCCTCACACATTGCTTGAATAAATTGGCTGAATCAGCTGTGCACTGCATCCGTTTTCTCCGAGGACTGTGTGTTGTAGCTTGGTCCCAGGGAATCCATCATGATCAAGGGAATAGTTGGCCTG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Stojkovic et al.
Journal of thrombosis and haemostasis : JTH, 12(6), 948-957 (2014-04-08)
Urokinase-type plasminogen activator (u-PA) plays a pivotal role in extracellular proteolysis and is thought to be critically involved in the modulation of angiogenesis. Interleukin (IL)-33 is a member of the IL-1 cytokine family, which is thought to act as danger
Xi-Xiang Yu et al.
Digestive diseases and sciences, 60(5), 1265-1272 (2015-02-07)
As a pro-inflammatory cytokine, IL-33 has been demonstrated to play an important role in tumor progression. It is reported that IL-33 is highly expressed in the serum and tumor tissues of patients with gastric cancer. However, the function of IL-33
Anne Marie Thompson et al.
American journal of physiology. Heart and circulatory physiology, 307(4), H533-H541 (2014-06-29)
Loss of vascular smooth muscle cell (VSMC) function is a hallmark of vascular disease. VSMCs become increasingly dysregulated, apoptotic, and senescent as we age. Sirtuin 1 (SirT1) is a deactylase that regulates substrates associated with stress mitigation, metabolism, and aging.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service