Skip to Content
Merck
All Photos(1)

Key Documents

EMU033001

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Gja3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCTTCGAAGTGGGGTTCATCGCGGGCCAGTACTTTCTATACGGCTTCCAGCTGCAGCCACTTTACCGCTGCGACCGCTGGCCCTGCCCCAACACTGTGGACTGTTTCATCTCCAGGCCCACAGAGAAGACCATCTTTGTCATCTTCATGCTGGCTGTGGCCTGTGCGTCACTGGTACTCAACATGCTGGAGATTTACCACCTGGGCTGGAAGAAGCTCAAGCAGGGAGTTACTAACCACTTCAACCCAGATGCCTCAGAAGCCAGGCACAAGCCCTTGGACCCCCTACCCACGGCCACCAGCTCTGGCCCGCCCAGCGTCTCCATCGGGTTCCCACCTTATTACACACACCCTGCCTGTCCCACAGTACAGGCAAAGGCCATAGGGTTTCCTGGGGCCCCACTATCACCAGCAGACTTCACAGTGGTGACTCTAAACGATGCTCAAGGCAGAAACCACCCAGTCAAACACTGCAATGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetsuya Muto et al.
Investigative ophthalmology & visual science, 55(7), 4327-4337 (2014-06-19)
To investigate whether high glucose (HG) alters connexin 43 (Cx43) expression and gap junction intercellular communication (GJIC) activity in retinal Müller cells, and promotes Müller cell and pericyte loss. Retinal Müller cells (rMC-1) and cocultures of rMC-1 and retinal pericytes
Tobias Forster et al.
Oncotarget, 5(6), 1621-1634 (2014-04-20)
The extreme aggressiveness of pancreatic ductal adenocarcinoma (PDA) has been associated with blocked gap junctional intercellular communication (GJIC) and the presence of cancer stem cells (CSCs). We examined whether disturbed GJIC is responsible for a CSC phenotype in established and
Dominique Thuringer et al.
Oncotarget, 6(12), 10267-10283 (2015-04-15)
High levels of circulating heat shock protein 70 (HSP70) are detected in many cancers. In order to explore the effects of extracellular HSP70 on human microvascular endothelial cells (HMEC), we initially used gap-FRAP technique. Extracellular human HSP70 (rhHSP70), but not
Lingzhi Wang et al.
Oncology reports, 34(4), 2133-2141 (2015-08-12)
Cisplatin, an important chemotherapeutic agent against testicular germ cell cancer, induces testicular toxicity on Leydig and Sertoli cells, leading to serious side-effects such as azoospermia and infertility. In a previous study, it was found that simvastatin enhanced the sensitivity of
S Morel et al.
Thrombosis and haemostasis, 112(2), 390-401 (2014-05-16)
Ubiquitous reduction of the gap junction protein Connexin43 (Cx43) in mice provides beneficial effects on progression and composition of atherosclerotic lesions. Cx43 is expressed in multiple atheroma-associated cells but its function in each cell type is not known. To examine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service