Skip to Content
Merck

Skip To

EMU021981

MISSION® esiRNA

targeting mouse Slc22a3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 212,00
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
€ 382,00

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324

€ 212,00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCATCAGTGTGGCAGAAAAAAGAAAACCCAGGTTTCTACTTCTGATGTCTGAGGCCCCTGTGTTGCCTGGAGGACCAGAGAAGACTGGAATATGCCTGTGCAGTGTGCTTCGCCTCCTCTAGGTGGCGCCCTTCCAAGGAACCGTAAAGATTTCGGAGTATTTTATAAGGTATAAGATGAAGATTTATCATGCTGCCCAAGCTTCGGGGAACACATAATCTTGGACCTGTTTCATGAAGACCCAGTAGGTACCCGGATTCTGGCTAGGTTCTCATTCAGCAAGTCTCTCACCCATTAGGTAAGGAGAGATGAAAAGGCCCCAACCAGTTCTCCAGTTCTCAGGACAAGACAGTACCATCCAGGAAGCAGGATGAACAAACCAAGGCCTGGCACGTGTCCAGAAGTGCGCTCAGCTGATCTGATCAACACACCGCAGTGAAGAGTG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMU021141EMU005341EMU088681
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

GCATCAGTGTGGCAGAAAAAAGAAAACCCAGGTTTCTACTTCTGATGTCTGAGGCCCCTGTGTTGCCTGGAGGACCAGAGAAGACTGGAATATGCCTGTGCAGTGTGCTTCGCCTCCTCTAGGTGGCGCCCTTCCAAGGAACCGTAAAGATTTCGGAGTATTTTATAAGGTATAAGATGAAGATTTATCATGCTGCCCAAGCTTCGGGGAACACATAATCTTGGACCTGTTTCATGAAGACCCAGTAGGTACCCGGATTCTGGCTAGGTTCTCATTCAGCAAGTCTCTCACCCATTAGGTAAGGAGAGATGAAAAGGCCCCAACCAGTTCTCCAGTTCTCAGGACAAGACAGTACCATCCAGGAAGCAGGATGAACAAACCAAGGCCTGGCACGTGTCCAGAAGTGCGCTCAGCTGATCTGATCAACACACCGCAGTGAAGAGTG

esiRNA cDNA target sequence

TGGAGCAAATTGCACAGAAGAACAGGAAGGTGCCCCCTGCTGACCTGAAGATGATGTGCCTTGAGGAAGATGCCTCAGAGAGGCGGAGTCCTTCGTTTGCAGACCTGTTCCGCACCCCCAGCCTGAGGAAGCACACCCTCATCCTGATGTATCTATGGTTCTCTTGTGCTGTGCTGTACCAGGGCCTCATCATGCATGTGGGAGCCACAGGGGCCAACCTCTACCTGGACTTCTTTTATTCTTCTCTAGTGGAATTCCCTGCGGCCTTTATCATCCTTGTCACCATTGACCGCATTGGCCGCATCTACCCAATAGCGGCATCAAATCTGGTGGCGGGGGCAGCCTGCCTCCTCATGATCTTTATCCCACATGAGCTGCATTGGTTGAATGTGACCCTCGCTTGTCTTGGCCGTATGGGAGCCACCATTGTGTTGCAGATGGTC

esiRNA cDNA target sequence

TCTTTGGCCTCACACTACCCAACTACCTGGATTCCAACCCAGGTGCTATCAACACAGGCATTCCTGAAGTGGATCAGATCCTAACTGTGCTGCTGACCACAGAGATGTTTGTTGGCGGGTGCCTTGCTTTCATACTGGACAACACAGTGCCAGGGAGCCCAGAGGAAAGAGGCCTGATACAGTGGAAGGCTGGAGCCCACGCCAACAGTGAGACGTCAGCCAGTCTCAAGAGCTACGATTTCCCCTTTGGGATGGGCATGGTAAAAAGGACCACCTTTTTCAGATACATCCCCATCTGCCCAGTCTTCAGAGGATTTTCTAAAAAGACACAAAATCAGCCTCCAGTTCTAGAAGACACTCCAGACAATATAGAAACTGGGTCTGTGTGTACCAAGGTCTGAAGCCACC

esiRNA cDNA target sequence

TGGGCCAGATCTCCAACTACGTGGTGGCCTTCATACTAGGAACTGAAATCCTGAGCAAGTCGGTTCGCATCATCTTCTCCACATTAGGAGTCTGTACATTTTTTGCAATCGGCTACATGGTCCTGCCGCTGTTTGCATACTTCATCAGAGACTGGAGGATGCTGCTGCTGGCCCTGACACTGCCTGGCCTGTTCTGTGTTCCCCTGTGGTGGTTTATTCCAGAATCTCCCCGGTGGCTGATATCCCAGAGGAGATTTGCAGAGGCCGAACAGATCATCCAGAAAGCCGCAAAGATGAACAGCATCGTGGCGCCAGCAGGGATATTCGATCCTCTAGAGCTACAGGAGCTAAACTCCTTGAAGCAGCAGAAAGTCATAATCCTGGACCTGTTCAGGACTCGGAACATTGCCACCATAACCGTGATGGCTGTGATGCTGTGGATGCTAACCTCAGTGGGTTACTTTGCTCTGTCTCTCAATGTTCCTAATTTACATGGAGATGTCTACCTGAACTGCTTCCTCTCTGGCCTGATTG

Gene Information

mouse ... SLC22A3(20519), Slc22a3(20519)

Gene Information

mouse ... SLC22A1(20517), Slc22a1(20517)

Gene Information

mouse ... SLC23A1(20522), Slc23a1(20522)

Gene Information

mouse ... SLC22A4(30805), Slc22a4(30805)

Ensembl | mouse accession no.

ENSMUSG00000023828

Ensembl | mouse accession no.

ENSMUSG00000023829

Ensembl | mouse accession no.

ENSMUSG00000024354

Ensembl | mouse accession no.

ENSMUSG00000020334

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

NCBI accession no.

NM_011395

NCBI accession no.

NM_009202

NCBI accession no.

NM_011397

NCBI accession no.

NM_019687


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters