Skip to Content
Merck
All Photos(1)

Key Documents

EMU021961

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Mapk14

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAGTGGAAGAGCCTGACCTATGATGAAGTCATCAGCTTTGTGCCACCACCCCTTGACCAAGAAGAAATGGAGTCCTGAGCACCTGGTTTCTGTTCTGTCTATCTCACTTCACTGTGAGGGGAAGACCTTCTCATGGGAACTCTCCAAATACCATTCAAGTGCCTCTTGTTGAAAGATTCCTTCATGGTGGAAGGGGGTGCATGTATGTGTTAGTGTTTGTGTGTGTGTGTGTGTCTGTCTGTTCGTCTGTCCACCTATCTTTGTGGAAGTCACTGTGATGGTAGTGACTTTATGAGTTGTGAATGGTCCTTGGCAGTCTGCCTGCTTTCTCAGAGTCTGGGCAGGCCGATGGGAACTGTCATCTCCTTAGGGATGTGTGTGTTCAGTGCAAAGTAAGAAATATGAAAATATCCCTGTTCTTAGTTACCTTGCCACTTTGGCTTC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hye Jeong Lee et al.
Molecular endocrinology (Baltimore, Md.), 29(6), 873-881 (2015-04-01)
Irisin is a novel myokine produced by skeletal muscle. However, its metabolic role is poorly understood. In the present study, irisin induced glucose uptake in differentiated skeletal muscle cells. It increased AMP-activated protein kinase (AMPK) phosphorylation and the inhibition of
Ana M Tormos et al.
PloS one, 12(2), e0171738-e0171738 (2017-02-07)
Hepatocyte poliploidization is an age-dependent process, being cytokinesis failure the main mechanism of polyploid hepatocyte formation. Our aim was to study the role of p38α MAPK in the regulation of actin cytoskeleton and cytokinesis in hepatocytes during development and aging.
Xiaoling Gu et al.
Free radical biology & medicine, 83, 149-158 (2015-03-17)
An increasing number of studies have focused on the phenomenon that mitochondrial DNA (mtDNA) activates innate immunity responses. However, the specific role of mtDNA in inflammatory lung disease remains elusive. This study was designed to examine the proinflammatory effects of
Desheng Zhong et al.
Journal of cellular biochemistry, 115(9), 1624-1635 (2014-05-03)
Pan-Bcl-2 family inhibitor obatoclax has been demonstrated to be effective against various cancers, of which the mechanism of action is not fully understood. In this study, we demonstrate that obatoclax suppressed esophageal cancer cell viability with concomitant G1/G0-phase cell cycle
Xi-Xi Lin et al.
Toxicology mechanisms and methods, 24(8), 575-583 (2014-08-20)
Cigarette smoke contains reactive oxygen (ROS) that can cause oxidative stress. It increases the number of apoptotic and necrotic lung cells and further induces the development of chronic airway disease. In this study, we investigated the effects of cigarette smoke

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service