Skip to Content
Merck
All Photos(1)

Key Documents

EMU016791

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Prmt1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGATGGGTTACTGCCTCTTCTACGAGTCCATGCTCAACACCGTGCTGCACGCTCGGGACAAGTGGCTGGCACCCGATGGCCTCATCTTCCCAGACCGGGCCACCTTGTATGTGACAGCCATTGAGGACCGACAATATAAAGACTACAAGATCCACTGGTGGGAGAACGTGTATGGCTTTGATATGTCCTGCATTAAAGACGTGGCCATCAAGGAGCCCCTGGTGGACGTGGTGGACCCAAAGCAGCTGGTCACCAATGCCTGCCTCATAAAGGAGGTGGACATTTACACAGTCAAGGTGGAGGACCTGACCTTCACCTCCCCCTTCTGCCTGCAAGTGAAGAGGAACGACTACGTGCACGCGCTGGTGGCTTACTTCAACATCGAGTTCACCCGATGCCACAAGAGGACCGGCTTCTCCACCAGTCCTGAG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sreedevi Avasarala et al.
The Journal of biological chemistry, 290(21), 13479-13489 (2015-04-08)
Protein arginine methyl transferase 1 (PRMT1) was shown to be up-regulated in cancers and important for cancer cell proliferation. However, the role of PRMT1 in lung cancer progression and metastasis remains incompletely understood. In the present study, we show that
Bingshou Li et al.
Biochemical and biophysical research communications, 464(4), 982-987 (2015-07-15)
Accumulating evidence indicates that microRNAs function as oncogenes or tumor suppressor genes in human cancer. MiR-503 is deregulated in various human cancers and has been associated with hepatocellular carcinoma (HCC) progression. However, the underlying mechanisms of miR-503 involvement in the
Dong-Il Kim et al.
Oxidative medicine and cellular longevity, 2015, 617919-617919 (2015-11-20)
Oxidative stress-induced retinal pigment epithelial (RPE) cell damage is involved in the progression of diabetic retinopathy. Arginine methylation catalyzed by protein arginine methyltransferases (PRMTs) has emerged as an important histone modification involved in diverse diseases. Sirtuin (SIRT1) is a protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service