Skip to Content
Merck
All Photos(1)

Key Documents

EMU013051

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Cav1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATCAGCACGCAGAAAGAGATATGAGGGACATTTCAAGGATGAAAGGTTTTTTCCCCCCTTACTATTTCCTTGGTGCCAATTCCAAGTTGCTCTCGCAGCAGCAAATTTATGAATGGTTTGTCTTGATCAAGAACAAAGAATTCATTCCCACCATTCTCATATATACTACTTGTCTCTTCTAAGCTACTGCATCTATGTTTGACAGTCTGGAATGTTTAAACCCATTCCTGCTCTCTCTTTTATATGTGAATCATTGTTTCATTGGCTAAAATATAAACATATTGTTGAAAGATGATTTGAGAAAAATAGGAAGGACTGGGAGGCAGGGAAGAGTACCAACAACCTCAACTGCCTACTCAAAGGTGATGATGTCATACAAAGGGAAGAGATTCAGGTTACGGCCATTTGTTTAGGGGCATGAAGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen Juks et al.
Biochimica et biophysica acta, 1848(12), 3205-3216 (2015-09-27)
Cell penetrating peptides are efficient tools to deliver various bioactive cargos into cells, but their exact functioning mechanism is still debated. Recently, we showed that a delivery peptide PepFect14 condenses oligonucleotides (ON) into negatively charged nanocomplexes that are taken up
Jing Zeng et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 37(4), 1289-1300 (2015-10-03)
Pulmonary microvascular endothelial cell (PMVEC) proliferation and angiogenesis contribute to the development of hepatopulmonary syndrome (HPS). MicroRNA-199a-5p (miR-199a-5p) has emerged as a potent regulator of angiogenesis, and its expression levels significantly decrease in the serum of patients with hepatopathy. However
Emily J Guggenheim et al.
Nanotoxicology, 14(4), 504-532 (2020-02-11)
Engineered Nanomaterials (NMs), such as Superparamagnetic Iron Oxide Nanoparticles (SPIONs), offer significant benefits in a wide range of applications, including cancer diagnostic and therapeutic strategies. However, the use of NMs in biomedicine raises safety concerns due to lack of knowledge
Fanrui Meng et al.
PloS one, 10(5), e0126056-e0126056 (2015-05-06)
Expression of Caveolin-1 (Cav1), a key component of cell surface caveolae, is elevated in prostate cancer (PCa) and associated with PCa metastasis and a poor prognosis for PCa patients. Polymerase I and Transcript Release Factor (PTRF)/cavin-1 is a cytoplasmic protein
Q Chen et al.
Cellular and molecular biology (Noisy-le-Grand, France), 61(2), 33-38 (2015-05-31)
Bladder cancer occurs in the majority of cases in males, which represents the fourth highest incident cancer in men and tenth in women. It is associated with a high rate of recurrence, and prognosis is poor once the cancer metastasizes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service