Skip to Content
Merck
All Photos(1)

Key Documents

EHU151901

Sigma-Aldrich

MISSION® esiRNA

targeting human ZEB1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATGGTGCAAGCTGTTGTTCTGCCAACAGTTGGTTTGGTGTCTCCCATAAGTATCAATTTAAGTGATATTCAGAATGTACTTAAAGTGGCGGTAGATGGTAATGTAATAAGGCAAGTGTTGGAGAATAATCAAGCCAATCTTGCATCCAAAGAACAAGAAACAATCAATGCTTCACCCATACAACAAGGTGGCCATTCTGTTATTTCAGCCATCAGTCTTCCTTTGGTTGATCAAGATGGAACAACCAAAATTATCATCAACTACAGTCTTGAGCAGCCTAGCCAACTTCAAGTTGTTCCTCAAAATTTAAAAAAAGAAAATCCAGTCGCTACAAACAGTTGTAAAAGTGAAAAGTTACCAGAAGATCTTACTGTTAAGTCTGAGAAGGACAAAAGCTTTGAAGGGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Jiang et al.
OncoTargets and therapy, 12, 6093-6104 (2019-08-24)
Objective: Gastric cancer (GC) is a common tumor malignancy with high incidence and poor prognosis. Radiotherapy is one of the main strategies for GC treatment, while development of radioresistance limits the effectiveness. microRNA-203 (miR-203) has been reported to participate in
Guanlin Wu et al.
American journal of translational research, 9(8), 3599-3610 (2017-09-02)
Epigenetic gene inactivation by microRNAs (miRNAs) is crucial in malignant transformation, prevention of apoptosis, development of drug resistance, and metastasis. miR-204 dysregulation has been reported in prostate cancer (PC). It is considered to exert tumor suppressor functions and is associated
Qiongyan Zou et al.
Journal of cellular biochemistry, 119(2), 2189-2199 (2017-09-01)
Breast cancer (BC) is one of the leading causes of cancer deaths worldwide and the most common cancer among women. In our previous study, we revealed that lncRNA TP73-AS1 promotes breast cancer cell proliferation through directly binding to miR-200a. Herein
Hong-Yan Zhang et al.
Gene, 633, 61-65 (2017-08-28)
The myocardial infarction associated transcript (MIAT), a long non-coding RNA (lncRNA), was originally identified as a candidate gene for myocardial infarction, and was recently shown to participate in the progression of cancer and the process of metastasis. However, the biological
Weili Liang et al.
Molecular medicine reports, 17(5), 6711-6716 (2018-03-08)
Breast cancer is a common malignant tumor in women. It has been suggested that a type of microcirculation pattern that does not rely on host microvascular endothelial cells known as vasculogenic mimicry (VM) may contribute to the poor effect of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service