Skip to Content
Merck
All Photos(1)

Key Documents

EHU151571

Sigma-Aldrich

MISSION® esiRNA

targeting human TWIST1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGGACAAGCTGAGCAAGATTCAGACCCTCAAGCTGGCGGCCAGGTACATCGACTTCCTCTACCAGGTCCTCCAGAGCGACGAGCTGGACTCCAAGATGGCAAGCTGCAGCTATGTGGCTCACGAGCGGCTCAGCTACGCCTTCTCGGTCTGGAGGATGGAGGGGGCCTGGTCCATGTCCGCGTCCCACTAGCAGGCGGAGCCCCCCACCCCCTCAGCAGGGCCGGAGACCTAGATGTCATTGTTTCCAGAGAAGGAGAAAATGGACAGTCTAGAGACTCTGGAGCTGGATAACTAAAAATAAAAATATATGCCAAAGATTTTCTTGGAAATTAGAAGAGCAAAATCCAAATTCAAAGAAACAGGGCGTGGGGCGCACTTTTAAAAGAGAAAGCGAGACAGGCCCGTGGACAGTGATTCCCAGACGGGCAGCGGCACCATCCTCACACCTCTGCATTCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanbiao Zhang et al.
Molecular medicine reports, 23(5) (2021-03-25)
Long non‑coding RNAs are associated with cancer progression. Long intergenic non‑protein coding RNA (linc)‑regulator of reprogramming (ROR) enhances tumor development in hepatocellular carcinoma (HCC). However, the effect of chemoresistance and its underlying mechanisms in HCC are not completely understood. The
Wei Li et al.
Oncology reports, 37(1), 185-192 (2016-11-24)
High expression of high mobility group protein A2 (HMGA2) is correlated with the invasiveness of gastric cancer and is an independent prognostic factor. The reason may be that HMGA2 promotes epithelial-mesenchymal transition (EMT) and the acquisition of tumor stem cell properties, yet
Yutian Wang et al.
Frontiers in microbiology, 11, 1301-1301 (2020-07-01)
Staphylococcus aureus (S. aureus) infection-induced osteomyelitis is a great challenge in clinic treatment. Identification of the essential genes and biological processes that are specifically changed in mononuclear cells at an early stage of S. aureus osteomyelitis is of great clinical
Mairéad A Cleary et al.
Stem cells and development, 26(10), 751-761 (2017-03-17)
Human bone marrow-derived mesenchymal stem cells (BMSCs) are clinically promising to repair damaged articular cartilage. This study investigated TWIST1, an important transcriptional regulator in mesenchymal lineages, in BMSC chondrogenesis. We hypothesized that downregulation of TWIST1 expression is required for in
Shi Chen et al.
Cancer letters, 383(1), 73-84 (2016-10-30)
The epithelial-mesenchymal transition (EMT) plays a crucial role in pancreatic ductal adenocarcinoma (PDAC) development and progression. TWIST activated by intra-tumoral hypoxia functions to promote the EMT. We hypothesized that TWIST and the downstream gene pathway could mediate PDAC progression under

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service