Skip to Content
Merck
All Photos(1)

Key Documents

EHU150081

Sigma-Aldrich

MISSION® esiRNA

targeting human SESN2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCTGTGCTTTGTGGAAGACCCTACTTTCGGATATGAGGACTTCACTCGGAGAGGGGCTCAGGCACCCCCTACCTTCCGGGCCCAGGATTATACCTGGGAAGACCATGGCTACTCGCTGATCCAGCGGCTTTACCCTGAGGGTGGGCAGCTGCTGGATGAGAAGTTCCAGGCAGCCTATAGCCTCACCTACAATACCATCGCCATGCACAGTGGTGTGGACACCTCCGTGCTCCGCAGGGCCATCTGGAACTATATCCACTGCGTCTTTGGCATCAGATATGATGACTATGATTATGGGGAGGTGAACCAGCTCCTGGAGCGGAACCTCAAGGTCTATATCAAGACAGTGGCCTGCTACCCAGAGAAGACCACCCGAAGAATGTACAACCTCTTCTGGAGGCACTTCCGCCACTCAGAGAAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Biochem/physiol Actions

SESN2 (sestrin-2) is a downstream effector of p53. It is associated with cell survival, protection and regeneration. It also plays an important role in autophagy induction and tumor suppression. Overexpression of SENS2 suppresses cancer growth.[1] It controls cell growth and proliferation by suppressing mTORC1 (mammalian target of rapamycin complex 1) activity through an AMPK (5′ AMP-activated protein kinase)-associated mechanism.[2] It is an antioxidant protein, and can be induced by oxidative and energetic stress.[3]

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An ShRNA Based Genetic Screen Identified Sesn2 as a Potential Tumor Suppressor in Lung Cancer via Suppression of Akt-mTOR-p70S6K Signaling.
Xu H
PLoS ONE, 10, e0124033-e0124033 (2015)
SESN2 correlates with advantageous prognosis in hepatocellular carcinoma.
Chen S
Diagnostic Pathology, 12, 13-13 (2017)
Sestrin2: A Promising Therapeutic Target for Liver Diseases.
Kim KM
Biological & Pharmaceutical Bulletin, 38, 966-966 (2015)
Mengjiao Chen et al.
Journal of cellular physiology, 236(1), 392-404 (2020-06-11)
Sestrin2 (SESN2) is a highly conservative oxidative stress protein that can regulate energy metabolism, cell proliferation, apoptosis, and mitochondria autophagy processes. It plays a role as an antioxidant in various diseases. The aims of the present study were to explore
Russell E Ericksen et al.
Cell metabolism, 29(5), 1151-1165 (2019-01-22)
Tumors display profound changes in cellular metabolism, yet how these changes aid the development and growth of tumors is not fully understood. Here we use a multi-omic approach to examine liver carcinogenesis and regeneration, and find that progressive loss of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service