Skip to Content
Merck
All Photos(1)

Documents

EHU149771

Sigma-Aldrich

MISSION® esiRNA

targeting human T

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGTCTCCTTCAGCAAAGTCAAGCTCACCAACAAGCTCAACGGAGGGGGCCAGATCATGCTGAACTCCTTGCATAAGTATGAGCCTCGAATCCACATAGTGAGAGTTGGGGGTCCACAGCGCATGATCACCAGCCACTGCTTCCCTGAGACCCAGTTCATAGCGGTGACTGCTTATCAGAACGAGGAGATCACAGCTCTTAAAATTAAGTACAATCCATTTGCAAAAGCTTTCCTTGATGCAAAGGAAAGAAGTGATCACAAAGAGATGATGGAGGAACCCGGAGACAGCCAGCAACCTGGGTACTCCCAATGGGGGTGGCTTCTTCCTGGAACCAGCACCCTGTGTCCACCTGCAAATCCTCATCCTCAGTTTGGAGGTGCCCTCTCCCTCCCCTCCACGCACAGCTGTGACAGGTACCCAACCCTGAGGAGCCACCGGTCCTCACCCTACCCCAGCCCCTATGCTCATCGGAACAATTCTCCAACCTATTCTGACAACTCACCTGCATGTTTATCCATGCTGCAATCCCATGACAATTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... T(6862) , T(6862)

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Hui Yu et al.
International journal of clinical and experimental pathology, 8(3), 2582-2589 (2015-06-06)
The aim of this study was to determine whether long non-coding RNA PVT1 can participate in the regulation of cardiac hypertrophy. A C57BL/6 mouse cardiac hypertrophic model was established using transverse aortic constriction (TAC). The animals subjected to sham operation
Zewen Kelvin Tuong et al.
PloS one, 11(1), e0147179-e0147179 (2016-01-27)
Nuclear hormone receptors have important roles in the regulation of metabolic and inflammatory pathways. The retinoid-related orphan receptor alpha (Rorα)-deficient staggerer (sg/sg) mice display several phenotypes indicative of aberrant lipid metabolism, including dyslipidemia, and increased susceptibility to atherosclerosis. In this
Jiaxuan Chen et al.
Journal of tissue engineering and regenerative medicine, 10(1), 40-51 (2013-06-21)
1α,25-Dihydroxyvitamin D3 [1α,25(OH)2D3] and bone morphogenetic protein-2 (BMP2) are both used to stimulate osteoblastic differentiation. 1α,25(OH)2D3 regulates osteoblasts through classical steroid hormone receptor mechanisms and through rapid responses that are mediated by two receptors, the traditional vitamin D receptor (VDR)
Y J Yang et al.
Cell death & disease, 6, e2025-e2025 (2015-12-18)
Taurine, which is found at high concentration in the heart, exerts several protective actions on myocardium. Physically, the high level of taurine in heart is maintained by a taurine transporter (TauT), the expression of which is suppressed under ischemic insult.
Alexander Michael Tabony et al.
Skeletal muscle, 4, 20-20 (2015-01-28)
Circulating angiotensin II (AngII) is elevated in congestive heart failure (CHF), and leads to skeletal muscle wasting, which is strongly associated with poor patient outcomes. We previously found that AngII upregulates protein phosphatase 2C-alpha (PP2Cα) and dephosphorylates AMP-activated protein kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service