Skip to Content
Merck
All Photos(1)

Key Documents

EHU148801

Sigma-Aldrich

MISSION® esiRNA

targeting human YBX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAGATGAGACCCAAGGTCAGCAGCCACCTCAACGTCGGTACCGCCGCAACTTCAATTACCGACGCAGACGCCCAGAAAACCCTAAACCACAAGATGGCAAAGAGACAAAAGCAGCCGATCCACCAGCTGAGAATTCGTCCGCTCCCGAGGCTGAGCAGGGCGGGGCTGAGTAAATGCCGGCTTACCATCTCTACCATCATCCGGTTTAGTCATCCAACAAGAAGAAATATGAAATTCCAGCAATAAGAAATGAACAAAAGATTGGAGCTGAAGACCTAAAGTGCTTGCTTTTTGCCCGTTGACCAGATAAATAGAACTATCTGCATTATCTATGCAGCATGGGGTTTTTATTATTTTTACCTAAAGACGTCTCTTTTTGGTAATAACAAACGTGTTTTTTAAAAAAGCCTGGTTTTTCTCAATACGCCTTTAAAGGTTTTTAAATTGTTTCATATCTGGTCAAGTTGAGATTTTTAAGAACTTCATTTTTAATTTGTAATAAAAGTTTACAACTTGATTTTTTCAAAAAAGTCAACAAACTGCAAGCACCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstanze Lettau et al.
International journal of radiation oncology, biology, physics, 109(2), 567-580 (2020-09-16)
Y-box binding protein 1 (YB-1) overexpression is associated with chemotherapy- and radiation therapy resistance. Ionizing radiation (IR), receptor tyrosine kinase ligands, and mutation in KRAS gene stimulate activation of YB-1. YB-1 accelerates the repair of IR-induced DNA double-strand breaks (DSBs).
Cristina Pagano et al.
Oncotarget, 8(4), 6193-6205 (2016-12-23)
Correct spatial and temporal control of cell proliferation is of fundamental importance for tissue homeostasis. Its deregulation has been associated with several pathological conditions. In common with almost every aspect of plant and animal biology, cell proliferation is dominated by
Aneri Shah et al.
Cancers, 12(8) (2020-08-09)
Cell fate decisions regulating survival and death are essential for maintaining tissue homeostasis; dysregulation thereof can lead to tumor development. In some cases, survival and death are triggered by the same receptor, e.g., tumor necrosis factor (TNF)-receptor 1 (TNFR1). We
Wei Wang et al.
Experimental and therapeutic medicine, 14(1), 499-506 (2017-07-05)
Antimicrobial peptide LL-37 serves a function in the host defense against microbial invasion, and also regulates cell proliferation, immune activity and angiogenesis. Previous studies have reported that LL-37 participates in the development of numerous tumour types, such as ovarian cancer
Takahisa Yamashita et al.
International journal of oncology, 51(2), 579-586 (2017-07-18)
The development and acquisition of multiple drug resistance in cancer cells remain a major obstacle in the treatment of bladder cancer. Nuclear translocation of Y box binding-1 (YB-1), which is a member of a family of DNA-binding proteins that contain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service