Skip to Content
Merck
All Photos(1)

Key Documents

EHU146141

Sigma-Aldrich

MISSION® esiRNA

targeting human RNF8

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGGACTTTGTCCTTTGTGGATTGCATAGCTGGATACCCATCATCTGTTTCTCTGATTGGAAGCTGCTGTTGTACAGAAAGACCTGCATTTCCCCCTTGTCTCCAGTTCTCTCACTACTTTTTCCTCCTCTGTGAGTGACCATCCAGGCAGTCACCATAACTGCTGGAGTGTCTGGGATTGGTAGCTCTCTCCAACTGCCTGCTTGCTCTTTACAGCCTCTCTCTGTGACTGGAATCTCTCCACCTCATCGTATCTAAGGATAACCCAGAAACATGGGGTGTCCTAGGTATGTTTATCTCGACACTGAACCCCCTAGGCTTCTGATGAATCCAGTGATTAGCTAAATTTGACATAGAAAGTAAGAAGGAATGTCTACTTTGTATTGTGGTCCTAATCTAAGATCAGGAGAATCCTGGAATTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Min et al.
Acta biochimica et biophysica Sinica, 51(8), 791-798 (2019-07-12)
MicroRNAs (miRNAs) are a class of endogenous noncoding genes that regulate gene expression at the posttranscriptional level. In recent decades, miRNAs have been reported to play important roles in tumor growth and metastasis, while some reported functions of a specific
Yoshiyuki Sasaki et al.
International journal of medical sciences, 10(9), 1231-1241 (2013-08-13)
The optimal timing of surgical resection of liver metastasis remains controversial, and guidelines regarding the upper limits of operative indications have not yet been defined. Surgical indication for metastasis from colorectal cancer (CLM) based on results of preoperative chemotherapy and
Shengli Wang et al.
Biochimica et biophysica acta, 1863(6), 1615-1628 (2017-02-22)
The ring finger protein 8 (RNF8), a key component of protein complex crucial for DNA-damage response, consists of a forkhead-associated (FHA) domain and a really interesting new gene (RING) domain that enables it to function as an E3 ubiquitin ligase.
Justine Sitz et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(39), 19552-19562 (2019-09-11)
High-risk human papillomaviruses (HR-HPVs) promote cervical cancer as well as a subset of anogenital and head and neck cancers. Due to their limited coding capacity, HPVs hijack the host cell's DNA replication and repair machineries to replicate their own genomes.
Maoxin Wang et al.
Oncology reports, 34(1), 341-349 (2015-05-09)
Tumor residue or recurrence is common after radiation therapy for nasopharyngeal cancer (NPC) since the tumor cells can repair irradiation-induced DNA damage. The ubiquitination cascade mediates the assembly of repair and signaling proteins at sites of DNA double-strand breaks (DSBs).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service