Skip to Content
Merck
All Photos(1)

Documents

EHU140981

Sigma-Aldrich

MISSION® esiRNA

targeting human MAPK10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAATCTTTCAGCCTGCTTCTTCCCCAGGGTTCTGTATTGCAGCTAAGCTCAAATGTATATTTAACTTCTAGTTGCTCTTGCTTTGGTCTTCTTCCAATGATGCTTACTACAGAAAGCAAATCAGACACAATTAGAGAAGCCTTTTCCATAAAGTGTAATTTTAATGGCTGCAAAACCGGCAACCTGTAACTGCCCTTTTAAATGGCATGACAAGGTGTGCAGTGGCCCCATCCAGCATGTGTGTGTCTCTATCTTGCATCTACCTGCTCCTTGGCCTAGTCAGATGGATGTAGATACAGATCCGCATGTGTCTGTATTCATACAGCACTACTTACTTAGAGATGCTACTGTCAGTGTCCTCAGGGCTCTACCAAGACATAATGCACTGGGGTACCACATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Wang et al.
Journal of medical genetics, 57(9), 634-642 (2020-02-19)
Hirschsprung disease (HSCR) is a life-threatening congenital disorder in which the enteric nervous system is completely missing from the distal gut. Recent studies have shown that miR-4516 markedly inhibits cell migration, and as one of its potential targets, MAPK10 functions
Baohua Qiao et al.
Reproductive sciences (Thousand Oaks, Calif.), 27(1), 380-388 (2020-02-13)
Ovarian cancer (OC) represents the most lethal form of gynaecologic cancers in developed countries. To develop a better therapeutic against OC, characterizing new classes of molecular regulators such as microRNAs (miRNAs) involved in OC tumorigenesis becomes immensely important. We used
Sarah Huntwork-Rodriguez et al.
The Journal of cell biology, 202(5), 747-763 (2013-08-28)
Neurons are highly polarized cells that often project axons a considerable distance. To respond to axonal damage, neurons must transmit a retrograde signal to the nucleus to enable a transcriptional stress response. Here we describe a mechanism by which this
Dan Ploug Christensen et al.
Journal of neuroinflammation, 13(1), 59-59 (2016-03-10)
Secretion of proteopathic α-synuclein (α-SNC) species from neurons is a suspected driving force in the propagation of Parkinson's disease (PD). We have previously implicated exophagy, the exocytosis of autophagosomes, as a dominant mechanism of α-SNC secretion in differentiated PC12 or
Lihua Li et al.
Oncology reports, 37(5), 2679-2687 (2017-04-11)
miRNA-27a-3p is an important regulator of carcinogenesis and other pathological processes. However, its role in laryngeal carcinoma is still unknown. In our previous research, we found that miR-27a-3p expression was upregulated in nasopharyngeal carcinoma (NPC) using a microarray chip. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service