Skip to Content
Merck
All Photos(1)

Key Documents

EHU136281

Sigma-Aldrich

MISSION® esiRNA

targeting human NPC1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGATTGTGGTGTTGGCTTTTGCCAAATCTCAAATTTTCCAGATATTCTACTTCAGGATGTATTTGGCCATGGTCTTACTGGGAGCCACTCACGGATTAATATTTCTCCCTGTCTTACTCAGTTACATAGGGCCATCAGTAAATAAAGCCAAAAGTTGTGCCACTGAAGAGCGATACAAAGGAACAGAGCGCGAACGGCTTCTAAATTTCTAGCCCTCTCGCAGGGCATCCTGACTGAACTGTGTCTAAGGGTCGGTCGGTTTACCACTGGACGGGTGCTGCATCGGCAAGGCCAAGTTGAACACCGGATGGTGCCAACCATCGGTTGTTTGGCAGCAGCTTTGAACGTAGCGCCTGTGAACTCAGGAATGCACAGTTGACTTGGGAAGCAGTATTACTAGATCTGGAGGCAACCACAGGACACTAAACTTCTCCCAGCCTCTTCAGGAAAGAAACCTCATTCTTTGGCAAGCAGGAGGTGACACTAGATGGCTGTGAATGTGATCCGCTCACTGACACTCTGTAAAGGCCAATCAATGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyang Xu et al.
Journal of cellular and molecular medicine, 21(2), 364-374 (2016-09-16)
Statins, 3-hydroxyl-3-methylglutaryl coenzyme A reductase inhibitors, are the first-line medications prescribed for the prevention and treatment of coronary artery diseases. The efficacy of statins has been attributed not only to their systemic cholesterol-lowering actions but also to their pleiotropic effects
Yudong Song et al.
Biomaterials, 150, 1-13 (2017-10-14)
Arginine and α-tocopherol succinate (α-TOS) double grafted N-trimethyl chitosan chloride (TMC) nanoparticles (TAS NPs) were designed and developed for effective co-delivery of doxorubicin (DOX) and Survivin shRNA-expressing pDNA (iSur-pDNA). With DOX loading into the hydrophobic core and iSur-pDNA combining to
Saskia Heybrock et al.
Nature communications, 10(1), 3521-3521 (2019-08-08)
The intracellular transport of cholesterol is subject to tight regulation. The structure of the lysosomal integral membrane protein type 2 (LIMP-2, also known as SCARB2) reveals a large cavity that traverses the molecule and resembles the cavity in SR-B1 that
Guanghong Liao et al.
Experimental neurology, 269, 67-74 (2015-04-14)
Niemann-Pick type C (NPC) disease is a genetic disorder associated with intracellular cholesterol accumulation in the brain and other organs, and neurodegeneration is generally believed to be the fatal cause of the disease. In view of the emerging role of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service