Skip to Content
Merck
All Photos(1)

Key Documents

EHU131771

Sigma-Aldrich

MISSION® esiRNA

targeting human HAS2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTGGGTGTGTTCAGTGCATTAGTGGACCTCTGGGAATGTACAGAAACTCCTTGTTGCATGAGTTTGTGGAAGATTGGTACAATCAAGAATTTATGGGCAACCAATGTAGCTTTGGTGATGACAGGCATCTCACGAACCGGGTGCTGAGCCTGGGCTATGCAACAAAATACACAGCTCGATCTAAGTGCCTTACTGAAACACCTATAGAATATCTCAGATGGCTAAACCAGCAGACCCGTTGGAGCAAGTCCTACTTCCGAGAATGGCTGTACAATGCAATGTGGTTTCACAAACATCACTTGTGGATGACCTACGAAGCGATTATCACTGGATTCTTTCCTTTCTTTCTCATTGCCACAGTAATCCAGCTCTTCTACCGGGGTAAAATTTGGAACATTCTCCTCTTCTTGTTAACTGTCCAGCTAGTAGGTCTCATAAAATCATCTTTTGCCAGCTGCCTTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Zhan et al.
Fertility and sterility, 114(4), 888-898 (2020-08-09)
To investigate the role(s) of hyaluronan synthase 2 (HAS2) and hyaluronan in disease progression of endometriosis and epidermal growth factor (EGF)-induced motility changes of endometriotic cells. A case-control experimental study and in vitro primary cell culture study. University hospital-affiliated research centers.
Xiaoyan Chen et al.
Cell communication and signaling : CCS, 18(1), 89-89 (2020-06-11)
Hyaluronan (HA) is an abundant component of the bone marrow (BM) extracellular matrix. Here, we investigated the abnormal deposition of HA in the BM microenvironment and its remodelling in mediating the malignancy of breast cancer cells (BCCs). BCCs were transplanted
Douglas Hanniford et al.
Cancer cell, 37(1), 55-70 (2020-01-15)
Metastasis is the primary cause of death of cancer patients. Dissecting mechanisms governing metastatic spread may uncover important tumor biology and/or yield promising therapeutic insights. Here, we investigated the role of circular RNAs (circRNA) in metastasis, using melanoma as a
Priscilla Soulié et al.
American journal of physiology. Cell physiology, 307(8), C745-C759 (2014-08-29)
Generation of branched tubes from an epithelial bud is a fundamental process in development. We hypothesized that induction of hyaluronan synthase (Has) and production of hyaluronan (HA) drives tubulogenesis in response to morphogenetic cytokines. Treatment of J3B1A mammary cells with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service