Skip to Content
Merck
All Photos(1)

Key Documents

EHU131661

Sigma-Aldrich

MISSION® esiRNA

targeting human ABCB1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCATGCTCAGACAGGATGTGAGTTGGTTTGATGACCCTAAAAACACCACTGGAGCATTGACTACCAGGCTCGCCAATGATGCTGCTCAAGTTAAAGGGGCTATAGGTTCCAGGCTTGCTGTAATTACCCAGAATATAGCAAATCTTGGGACAGGAATAATTATATCCTTCATCTATGGTTGGCAACTAACACTGTTACTCTTAGCAATTGTACCCATCATTGCAATAGCAGGAGTTGTTGAAATGAAAATGTTGTCTGGACAAGCACTGAAAGATAAGAAAGAACTAGAAGGTTCTGGGAAGATCGCTACTGAAGCAATAGAAAACTTCCGAACCGTTGTTTCTTTGACTCAGGAGCAGAAGTTTGAACATATGTATGCTCAGAGTTTGCAGGTACCATACAGAAACTCTTTGAGGAAAGCACACATCTTTGGAATTACATTTTCCTTCACCCAGGCAATGATGTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho Kyung Seo et al.
The Prostate, 80(6), 453-462 (2020-03-07)
Docetaxel is the preferred chemotherapeutic agent for hormone-refractory prostate cancer (PC) patients. However, patients eventually develop docetaxel resistance, and no effective treatment options are available for them. We aimed to establish docetaxel resistance in castration-resistant prostate cancer (CRPC) cell lines
Vlasta Němcová-Fürstová et al.
Toxicology and applied pharmacology, 310, 215-228 (2016-09-25)
Development of taxane resistance has become clinically very important issue. The molecular mechanisms underlying the resistance are still unclear. To address this issue, we established paclitaxel-resistant sublines of the SK-BR-3 and MCF-7 breast cancer cell lines that are capable of
Michael Jelínek et al.
Toxicology and applied pharmacology, 347, 79-91 (2018-04-07)
We tested the role of substituents at the C3' and C3'N positions of the taxane molecule to identify taxane derivatives capable of overcoming acquired resistance to paclitaxel. Paclitaxel-resistant sublines SK-BR-3/PacR and MCF-7/PacR as well as the original paclitaxel-sensitive breast cancer
Tao Wang et al.
Theranostics, 5(12), 1456-1472 (2015-12-19)
Understanding the molecular basis of drug resistance and utilising this information to overcome chemoresistance remains a key challenge in oncology. Here we report that survivin, a key protein implicated in drug resistance, is overexpressed in cancer stem cell pool of
Xiaoqian Yang et al.
Pharmaceutical research, 32(6), 2097-2109 (2014-12-18)
Approaches for the synthesis of biomaterials to facilitate the delivery of "biologics" is a major area of research in cancer therapy. Here we designed and characterized a hyaluronic acid (HA) based self-assembling nanoparticles that can target CD44 receptors overexpressed on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service