Skip to Content
Merck
All Photos(1)

Key Documents

EHU129311

Sigma-Aldrich

MISSION® esiRNA

targeting human ADAM10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGAACTCTGCCATTTCACTCTGTCATTTATCATGAAGATGATATTAACTATCCCCATAAATACGGTCCTCAGGGGGGCTGTGCAGATCATTCAGTATTTGAAAGAATGAGGAAATACCAGATGACTGGTGTAGAGGAAGTAACACAGATACCTCAAGAAGAACATGCTGCTAATGGTCCAGAACTTCTGAGGAAAAAACGTACAACTTCAGCTGAAAAAAATACTTGTCAGCTTTATATTCAGACTGATCATTTGTTCTTTAAATATTACGGAACACGAGAAGCTGTGATTGCCCAGATATCCAGTCATGTTAAAGCGATTGATACAATTTACCAGACCACAGACTTCTCCGGAATCCGTAACATCAGTTTCATGGTGAAACGCATAAGAATCAATACAACTGCTGATGAGAAGGACCCTACAAATCCTTTCCGTTTCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zikang Xie et al.
Environmental toxicology, 35(8), 867-878 (2020-03-22)
MiR-20a has been reported as a key regulator to pro-inflammatory factor release in fibroblast-like synoviocytes (FLS), which caused rheumatoid arthritis (RA). However, the molecular mechanism of miR-20a in RA remains to be further elucidated. This study aimed to investigate the
Xiufen Liu et al.
Communications biology, 3(1), 728-728 (2020-12-03)
Mesothelin (MSLN) is a lineage restricted cell surface protein expressed in about 30% of human cancers and high MSLN expression is associated with poor survival in several different cancers. The restricted expression of MSLN in normal tissue and its frequent
Quan Yuan et al.
Cancer cell international, 20, 99-99 (2020-04-08)
Osteosarcoma is a malignant bone tumor. Increasing evidences have revealed that a disintegrin and metalloproteinase 10 (ADAM10) is implicated in tumor development. The main purpose of this study is to explore the effects of ADAM10 on osteosarcoma cell functions and
Wen Wu et al.
Inflammation, 43(2), 673-685 (2019-12-18)
Aseptic loosening (AL) is the most frequent cause of failure of total hip arthroplasties (THA). Prosthetic wear particle-induced monocyte recruitment to the periprosthetic tissue and subsequent inflammatory response are thought to be the major contribution to AL. Fibroblast is a
Zhuo-peng Ye et al.
PloS one, 9(8), e105350-e105350 (2014-08-16)
CD8+ T cells play an important role in the anti-tumor activities of the body. The dysfunction of CD8+ T cells in glioma is unclear. This study aims to elucidate the glioma cell-derived ADAM10 (A Disintegrin and metalloproteinase domain-containing protein 10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service