Skip to Content
Merck
All Photos(1)

Key Documents

EHU127401

Sigma-Aldrich

MISSION® esiRNA

targeting human BRAF

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACCAGCAAGCTAGATGCACTCCAACAAAGAGAACAACAGTTATTGGAATCTCTGGGGAACGGAACTGATTTTTCTGTTTCTAGCTCTGCATCAATGGATACCGTTACATCTTCTTCCTCTTCTAGCCTTTCAGTGCTACCTTCATCTCTTTCAGTTTTTCAAAATCCCACAGATGTGGCACGGAGCAACCCCAAGTCACCACAAAAACCTATCGTTAGAGTCTTCCTGCCCAACAAACAGAGGACAGTGGTACCTGCAAGGTGTGGAGTTACAGTCCGAGACAGTCTAAAGAAAGCACTGATGATGAGAGGTCTAATCCCAGAGTGCTGTGCTGTTTACAGAATTCAGGATGGAGAGAAGAAACCAATTGGTTGGGACACTGATATTTCCTGGCTTACTGGAGAAGAATTGCATGTGGAAGTGTTGGAGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Zhang et al.
The EMBO journal, 38(15), e100871-e100871 (2019-07-16)
Reactive oxygen species (ROS) are emerging as important regulators of cancer growth and metastatic spread. However, how cells integrate redox signals to affect cancer progression is not fully understood. Mitochondria are cellular redox hubs, which are highly regulated by interactions
Samantha K McCarty et al.
Endocrine-related cancer, 21(6), 865-877 (2014-09-18)
Increased p21-activated kinase (PAK) signaling and expression have been identified in the invasive fronts of aggressive papillary thyroid cancers (PTCs), including those with RET/PTC, BRAFV600E, and mutant RAS expression. Functionally, thyroid cancer cell motility in vitro is dependent on group
Nicholes R Candelaria et al.
PloS one, 10(12), e0145061-e0145061 (2015-12-15)
Hereditary, hormonal, and behavioral factors contribute to the development of breast cancer. Alcohol consumption is a modifiable behavior that is linked to increased breast cancer risks and is associated with the development of hormone-dependent breast cancers as well as disease
Yujuan Zhang et al.
Nanomedicine : nanotechnology, biology, and medicine, 14(5), 1679-1693 (2018-04-24)
Melanoma is significantly associated with mutant BRAF gene, a suitable target for siRNA-based anti-melanoma therapy. However, a tumor-specific delivery system is a major hurdle for clinical applications. Here, we developed a novel nano-carrier, FA-GNR-siBRAF for safe topical application, which consists
Lin Feng et al.
Biochemical and biophysical research communications, 524(1), 28-35 (2020-01-26)
BRAFV600E mutation is frequently observed in melanoma, and contributes to tumor malignancy. Despite inhibition of BRAF causes a profound cell growth inhibition and a strong clinical benefit in BRAFV600E melanoma, acquired drug resistance is still the major hurdle. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service