Skip to Content
Merck
All Photos(1)

Documents

EHU123581

Sigma-Aldrich

MISSION® esiRNA

targeting human THPO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCTCAGACACTGCCGACATCAGCATTGTCTCGTGTACAGCTCCCTTCCCTGCAGGGCGCCCCTGGGAGACAACTGGACAAGATTTCCTACTTTCTCCTGAAACCCAAAGCCCTGGTAAAAGGGATACACAGGACTGAAAAGGGAATCATTTTTCACTGTACATTATAAACCTTCAGAAGCTATTTTTTTAAGCTATCAGCAATACTCATCAGAGCAGCTAGCTCTTTGGTCTATTTTCTGCAGAAATTTGCAACTCACTGATTCTCTACATGCTCTTTTTCTGTGATAACTCTGCAAAGGCCTGGGCTGGCCTGGCAGTTGAACAGAGGGAGAGACTAACCTTGAGTCAGAAAACAGAGAAAGGGTAATTTCCTTTGCTTCAAATTCAAGGCCTTCCAACGCCCCCATCCCCTTTACTATCATTCTCAGTGGGACTCTGATCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Zou et al.
Cancer biomarkers : section A of Disease markers, 25(2), 133-139 (2018-11-20)
Long noncoding RNAs (LncRNAs) are involved in the occurrence and progression of human tumors including ovarian cancer (OC). Long noncoding RNA HOTTIP has been found to be involved in several human tumors development. However, the role of HOTTIP in OC
Bo-Wen Dai et al.
Journal of Cancer, 10(19), 4540-4551 (2019-09-19)
As a master regulator of embryonic morphogenesis, homeodomain-containing gene 10 (HOXC10) has been found to promote progression of human cancers and indicate poor survival outcome. Therefore, we concentrate on elucidating the role of HOXC10 in progression of oral squamous cell
Hideo Otsuki et al.
The Prostate, 77(2), 222-233 (2016-10-04)
Leucine stimulates cancer cell proliferation through the mTOR pathway, therefore, inhibiting leucine transporters may be a novel therapeutic target for cancer. L-type amino acid transporter (LAT) 1, a Na LNCaP and DU145 and PC-3 cell lines were used as a
Bo Li et al.
Free radical research, 52(4), 390-401 (2018-02-06)
Substantial evidence indicates that the alteration of the cellular redox status is a critical factor involved in cell growth and death and results in tumourigenesis. Cancer cells have an efficient antioxidant system to counteract the increased generation of ROS. However
Lina M E Pettersson et al.
PloS one, 9(6), e100730-e100730 (2014-06-27)
Peripheral nerve injury results in dramatic upregulation in pituitary adenylate cyclase activating polypeptide (PACAP) expression in adult rat dorsal root ganglia and spinal motor neurons mirroring that described for the neurotrophin brain derived neurotrophic factor (BDNF). Thus, we posited that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service