Skip to Content
Merck
All Photos(1)

Key Documents

EHU120391

Sigma-Aldrich

MISSION® esiRNA

targeting human S100A6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAAAGTCGAGGGGGTAAACCGCGAATGTGCGTTGTGTAAGCCACGGCGCAGGGTGGGGCGCGGGCGGGACTTGGGCGGGCGGGGTGGGCTTGGCCGAGCTGGCCTCCGGGGCACCGACCGCTATAAGGCCAGTCGGACTGCGACACAGCCCATCCCCTCGACCGCTCGCGTCGCATTTGGCCGCCTCCCTACCGCTCCAAGCCCAGCCCTCAGCCATGGCATGCCCCCTGGATCAGGCCATTGGCCTCCTCGTGGCCATCTTCCACAAGTACTCCGGCAGGGAGGGTGACAAGCACACCCTGAGCAAGAAGGAGCTGAAGGAGCTGATCCAGAAGGAGCTCACCATTGGCTCGAAGCTGCAGGATGCTGAAATTGCAAGGCTGATGGAAGACTTGGACCGGAACAAGGACCAGGAGGTGAACTTCCAGGAGTATGTCACCTTCCTGGGGGCCTTGGCTTTGATCTACAATGAAGCCCTCAAGGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Tamai et al.
Blood cancer journal, 1(11), e38-e38 (2012-07-26)
Mixed-lineage leukemia (MLL)-AFF1 (MLL-AF4)-positive acute lymphoblastic leukemia (ALL) is associated with poor prognosis, even after allogeneic hematopoietic stem cell transplantation (allo-HSCT). The resistance to graft-versus-leukemia (GVL) effects may be responsible for the poor effect of allo-HSCT on MLL-AFF1-positive ALL. Cytotoxic
Yaoming Peng et al.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology, 34(9), 815-820 (2018-03-17)
S100 calcium-binding protein A6 (S100A6) is up-regulated in many malignancies and overexpression of S100A6 has been identified associated with proliferation, migration and invasion phenotype in several cancer cells. In the present study, we explored whether S100A6 plays a role in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service