Skip to Content
Merck
All Photos(1)

Key Documents

EHU092971

Sigma-Aldrich

MISSION® esiRNA

targeting human CD151

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AACACGGAGCTCAAGGAGAACCTGAAGGACACCATGACCAAGCGCTACCACCAGCCGGGCCATGAGGCTGTGACCAGCGCTGTGGACCAGCTGCAGCAGGAGTTCCACTGCTGTGGCAGCAACAACTCACAGGACTGGCGAGACAGTGAGTGGATCCGCTCACAGGAGGCCGGTGGCCGTGTGGTCCCAGACAGCTGCTGCAAGACGGTGGTGGCTCTTTGTGGGCAGCGAGACCATGCCTCCAACATCTACAAGGTGGAGGGCGGCTGCATCACCAAGTTGGAGACCTTCATCCAGGAGCACCTGAGGGTCATTGGGGCTGTGGGGATCGGCATTGCCTGTGTGCAGGTCTTTGGCATGATCTTCACGTGCTGCCTGTAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura A Fast et al.
International journal of molecular sciences, 19(10) (2018-10-04)
Tetraspanins are suggested to regulate the composition of cell membrane components and control intracellular transport, which leaves them vulnerable to utilization by pathogens such as human papillomaviruses (HPV) and cytomegaloviruses (HCMV) to facilitate host cell entry and subsequent infection. In
Yongkang Qiao et al.
The Journal of allergy and clinical immunology, 139(1), 82-92 (2016-05-29)
Airway smooth muscle (ASM) contraction underpins airway constriction; however, underlying mechanisms for airway hyperresponsiveness (AHR) remain incompletely defined. CD151, a 4-transmembrane glycoprotein that associates with laminin-binding integrins, is highly expressed in the human lung. The role of CD151 in ASM
Yongkang Qiao et al.
The Journal of allergy and clinical immunology, 141(5), 1799-1817 (2017-12-24)
Despite advances in our understanding of the mechanisms of influenza A virus (IAV) infection, the crucial virus-host interactions during the viral replication cycle still remain incomplete. Tetraspanin CD151 is highly expressed in the human respiratory tract, but its pathological role in
Jérôme Finke et al.
Scientific reports, 10(1), 5356-5356 (2020-03-27)
During cell invasion, human papillomaviruses use large CD151 patches on the cell surface. Here, we studied whether these patches are defined architectures with features for virus binding and/or internalization. Super-resolution microscopy reveals that the patches are assemblies of closely associated
Soonyean Hwang et al.
Cellular and molecular life sciences : CMLS, 76(8), 1595-1604 (2019-02-20)
Tetraspanin protein CD151 has typically been studied as binding partner and functional regulator of laminin-binding integrins. However, we show here that CD151 supports anti-cancer drug resistance independent of integrins. CD151 ablation sensitized multiple tumor cell types to several anti-cancer drugs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service