Skip to Content
Merck
All Photos(1)

Key Documents

EHU087831

Sigma-Aldrich

MISSION® esiRNA

targeting human TRIM29

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCATGGATGCTCTGGATGAGAGAGCCAAGGTGCTGCATGAGGACAAGCAGACCCGGGAGCAGCTGCATAGCATCAGCGACTCTGTGTTGTTTCTGCAGGAATTTGGTGCATTGATGAGCAATTACTCTCTCCCCCCACCCCTGCCCACCTATCATGTCCTGCTGGAGGGGGAGGGCCTGGGACAGTCACTAGGCAACTTCAAGGACGACCTGCTCAATGTATGCATGCGCCACGTTGAGAAGATGTGCAAGGCGGACCTGAGCCGTAACTTCATTGAGAGGAACCACATGGAGAACGGTGGTGACCATCGCTATGTGAACAACTACACGAACAGCTTCGGGGGTGAGTGGAGTGCACCGGACACCATGAAGAGATACTCCATGTACCTGACACCCAAAGGTGGGGTCCGGACATCATACCAGCCCTCGTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Han et al.
Aging, 13(4), 5034-5054 (2021-01-27)
Targeted molecular therapy is the most effective treatment for cancer. An effective therapeutic target for colorectal cancer (CRC) is urgently needed. However, the mechanisms of CRC remain poorly understood, which has hampered research and development of CRC-targeted therapy. TRIM29 is
Phillip L Palmbos et al.
Cancer research, 75(23), 5155-5166 (2015-10-17)
Bladder cancer is a common and deadly malignancy but its treatment has advanced little due to poor understanding of the factors and pathways that promote disease. ATDC/TRIM29 is a highly expressed gene in several lethal tumor types, including bladder tumors
Yasushi Masuda et al.
Nature communications, 6, 7299-7299 (2015-06-23)
Although DNA double-strand break (DSB) repair is mediated by numerous proteins accumulated at DSB sites, how DNA repair proteins are assembled into damaged chromatin has not been fully elucidated. Here we show that a member of the tripartite motif protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service