Skip to Content
Merck
All Photos(1)

Key Documents

EHU085701

Sigma-Aldrich

MISSION® esiRNA

targeting human APP

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTCGTTCCTGACAAGTGCAAATTCTTACACCAGGAGAGGATGGATGTTTGCGAAACTCATCTTCACTGGCACACCGTCGCCAAAGAGACATGCAGTGAGAAGAGTACCAACTTGCATGACTACGGCATGTTGCTGCCCTGCGGAATTGACAAGTTCCGAGGGGTAGAGTTTGTGTGTTGCCCACTGGCTGAAGAAAGTGACAATGTGGATTCTGCTGATGCGGAGGAGGATGACTCGGATGTCTGGTGGGGCGGAGCAGACACAGACTATGCAGATGGGAGTGAAGACAAAGTAGTAGAAGTAGCAGAGGAGGAAGAAGTGGCTGAGGTGGAAGAAGAAGAAGCCGATGATGACGAGGACGATGAGGATGGTGATGAGGTAGAGGAAGAGGCTGAGGAACCCTACGAAGAAGCCACAGAGAGAACCACCAGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... APP(351) , APP(351)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruce X Wong et al.
PloS one, 9(12), e114174-e114174 (2014-12-03)
Ceruloplasmin is a ferroxidase that interacts with ferroportin to export cellular iron, but is not expressed in neurons. We recently reported that the amyloid precursor protein (APP) is the analogous iron-exporting chaperone for neurons and other cells. The ferroxidase activity
Changyi Ji et al.
Cellular and molecular neurobiology, 38(4), 941-954 (2017-11-28)
Iron efflux in mammalian cells is mediated by the ferrous iron exporter ferroportin (Fpn); Fpn plasma membrane localization and function are supported by a multicopper ferroxidase and/or the soluble amyloid precursor protein (sAPP). Fpn and APP are ubiquitously expressed in
Nathalie Pierrot et al.
EMBO molecular medicine, 5(4), 608-625 (2013-04-05)
Perturbation of lipid metabolism favours progression of Alzheimer disease, in which processing of Amyloid Precursor Protein (APP) has important implications. APP cleavage is tightly regulated by cholesterol and APP fragments regulate lipid homeostasis. Here, we investigated whether up or down
Livius V d'Uscio et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 38(10), 1715-1726 (2017-09-30)
The exact physiological function of amyloid-β precursor protein (APP) in endothelial cells is unknown. Endothelium-specific APP-deficient (eAPP-/-) mice were created to gain new insights into the role of APP in the control of vascular endothelial function. Endothelium-dependent relaxations to acetylcholine
Philipp Spitzer et al.
Frontiers in immunology, 11, 1967-1967 (2020-10-06)
It has been previously shown that the amyloid precursor protein (APP) support the innate immune defense as an immune receptor. Amyloid β (Aβ) peptides seem to have properties of an antimicrobial peptide and can act as opsonines. In APP-deficient mouse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service