Skip to Content
Merck
All Photos(1)

Key Documents

EHU085291

Sigma-Aldrich

MISSION® esiRNA

targeting human JMJD6

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTTGTTGAATGCGCAAGAGGGCTGGTCTGCGCAGGAGAAATGGACTCTGGAGCGCCTAAAAAGGAAATATCGGAACCAGAAGTTCAAGTGTGGTGAGGATAACGATGGCTACTCAGTGAAGATGAAGATGAAATACTACATCGAGTACATGGAGAGCACTCGAGATGATAGTCCCCTTTACATCTTTGACAGCAGCTATGGTGAACACCCTAAAAGAAGGAAACTTTTGGAAGACTACAAGGTGCCAAAGTTTTTCACTGATGACCTTTTCCAGTATGCTGGGGAGAAGCGCAGGCCCCCTTACAGGTGGTTTGTGATGGGGCCACCACGCTCCGGAACTGGGATTCACATCGACCCTCTGGGAACCAGTGCCTGGAATGCCTTAGTTCAGGGCCACAAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang-Ryul Lee et al.
Carcinogenesis, 37(2), 119-128 (2015-12-10)
Cancer stem cells (CSCs) are defined as a small subpopulation of cancer cells within a tumor and responsible for initiation and maintenance of tumor growth. Thus, understanding of molecular regulators of CSCs is of paramount importance for the development of
Yan Liu et al.
Oncogene, 38(7), 980-997 (2018-09-07)
Overexpression of Jumonji domain-containing 6 (JMJD6) has been reported to be associated with more aggressive breast cancer characteristics. However, the precise role of JMJD6 in breast cancer development remains unclear. Here, we demonstrate that JMJD6 has intrinsic tyrosine kinase activity
Alec Paschalis et al.
Cancer research, 81(4), 1087-1100 (2021-04-07)
Endocrine resistance (EnR) in advanced prostate cancer is fatal. EnR can be mediated by androgen receptor (AR) splice variants, with AR splice variant 7 (AR-V7) arguably the most clinically important variant. In this study, we determined proteins key to generating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service