Skip to Content
Merck
All Photos(1)

Key Documents

EHU074691

Sigma-Aldrich

MISSION® esiRNA

targeting human GABPA

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAGAGTGCACAGAAGAAAGCATTGTAGAACAAACCTACGCGCCAGCTGAATGTGTAAGCCAGGCCATAGACATCAATGAACCAATAGGCAATTTAAAGAAACTGCTAGAACCAAGACTACAGTGTTCTTTGGATGCTCATGAAATTTGTCTGCAAGATATCCAGCTGGATCCAGAACGAAGTTTATTTGACCAAGGAGTAAAAACAGATGGAACTGTACAGCTTAGTGTACAGGTAATTTCTTACCAAGGAATTGAACCAAAGTTAAACATCCTTGAAATTGTTAAACCTGCGGACACTGTTGAGGTTGTTATTGATCCAGATGCCCACCATGCTGAATCAGAAGCACATCTTGTTGAAGAAGCTCAAGTGATAACTCTTGATGGCACAAAACACATCACAACCATTTCAGATGAAACTTCAGAACAAGTGACAAGATGGGCTGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Limin Xu et al.
Frontiers in cell and developmental biology, 8, 569977-569977 (2020-10-31)
Cerebral ischemic injury is a complicated pathological process. Adipose-derived stromal cells (ADSCs) have been used as a therapeutic strategy, with their therapeutic effects chiefly attributed to paracrine action rather than trans-differentiation. Studies have shown that circAkap7 was found to be
Lingjie Bao et al.
Molecular carcinogenesis, 56(6), 1543-1553 (2017-01-24)
Previously, we have demonstrated that NRF2 plays a key role in mediating cisplatin resistance in ovarian cancer. To further explore the mechanism underlying NRF2-dependent cisplatin resistance, we stably overexpressed or knocked down NRF2 in parental and cisplatin-resistant human ovarian cancer
Arpita Chatterjee et al.
Redox biology, 34, 101542-101542 (2020-05-04)
Radiation is a common anticancer therapy for many cancer patients, including prostate cancer. Diabetic prostate cancer patients suffer from increased lymph node metastasis, tumor recurrence and decreased survival as compared to non-diabetic prostate cancer patients. These patients are also at
Yanxia Guo et al.
Cell death and differentiation, 27(6), 1862-1877 (2019-12-06)
TERT promoter mutations occur in the majority of glioblastoma, bladder cancer (BC), and other malignancies while the ETS family transcription factors GABPA and its partner GABPB1 activate the mutant TERT promoter and telomerase in these tumors. GABPA depletion or the
Lu Gao et al.
Biochimica et biophysica acta. Molecular basis of disease, 1864(10), 3322-3338 (2018-07-22)
Diabetes contributes to cardiovascular complications and the pathogenesis of cardiac remodeling that can lead to heart failure. We aimed to evaluate the functional role of LAZ3 in diabetic cardiomyopathy (DCM). Streptozotocin (STZ) was used to induce a diabetic mouse model.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service