Skip to Content
Merck
All Photos(1)

Key Documents

EHU074341

Sigma-Aldrich

MISSION® esiRNA

targeting human OXA1L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AATGTGGGCTGTTCTTGAGCTAGGTGCTGAGACAGGTGTGCAAAGTTCTGACCTTCAGTGGATGAGAAATGTCATCAGAATGATGCCCCTGATAACCTTGCCCATAACCATGCATTTCCCCACGGCAGTGTTTATGTACTGGCTCTCCTCCAATTTGTTTTCCCTGGTCCAAGTATCCTGTCTCCGGATTCCAGCAGTACGCACTGTACTTAAAATCCCCCAGCGTGTTGTACATGACCTGGACAAATTACCTCCACGGGAAGGCTTCCTAGAGAGCTTCAAAAAAGGCTGGAAAAATGCTGAAATGACGCGTCAGCTGCGAGAGCGTGAACAACGCATGCGGAATCAGTTGGAGCTAGCAGCCAGGGGTCCTTTACGACAGACCTTTACCCACAACCCTCTCCTACAACCTGGAAAGGATAACCCTCCCAATATCCCTAGCAGCAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongfei Chen et al.
Bioengineered, 11(1), 1269-1279 (2020-11-04)
Emerging evidence suggested that circular RNAs (circRNAs) play critical roles in cervical cancer (CC) progression. However, the roles and molecular mechanisms of hsa_circ_0007364 in the tumorigenesis of CC remain unclear. In the present study, we used bioinformatics analysis and a
Yuhan Chen et al.
Gene, 629, 35-42 (2017-08-05)
Radiation-induced liver fibrosis (RILF) is considered as a major complication of radiation therapy for liver cancer. Circular RNA (circRNA) has been recently identified as a functional noncoding RNA involving in various biological processes. However, the expression pattern and regulatory capacity
Beibei Shao et al.
Biochemical and biophysical research communications, 513(1), 135-140 (2019-04-05)
Recent studies indicated that circular RNAs (circRNAs) could play critical roles in the initiation and development of tumors, including tongue squamous cell carcinoma (TSCC). We aimed to investigate the roles and underlying mechanisms of hsa_circ_0001742 in TSCC. In the present
Wei Liu et al.
Biochemical and biophysical research communications, 500(4), 846-851 (2018-04-27)
Lung cancer characterized with malignant cell growth is the leading cause of cancer-related deaths. In recent years, several circular RNAs (circRNAs) have been reported to participate in lung cancer progression. However, the correlation between circular RNA (circRNA) and lung cancer still
Jian Li et al.
Cell transplantation, 30, 963689720975390-963689720975390 (2021-01-14)
Gastric cancer (GC) is a big threat to human life and health. Circular RNAs (circRNAs), a subclass of noncoding RNAs, were reported to play a critical role in GC progression. Here, we investigated the role of a novel circRNA named

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service