Skip to Content
Merck
All Photos(1)

Documents

EHU070821

Sigma-Aldrich

MISSION® esiRNA

targeting human PDCD4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AATGAAGTTGCGGAAATGTTAAGAGATTTAAATCTTGGTGAAATGAAAAGTGGAGTACCAGTGTTGGCAGTATCCTTAGCATTGGAGGGGAAGGCTAGTCATAGAGAGATGACATCTAAGCTTCTTTCTGACCTTTGTGGGACAGTAATGAGCACAACTGATGTGGAAAAATCATTTGATAAATTGTTGAAAGATCTACCTGAATTAGCACTGGATACTCCTAGAGCACCACAGTTGGTGGGCCAGTTTATTGCTAGAGCTGTTGGAGATGGAATTTTATGTAATACCTATATTGATAGTTACAAAGGAACTGTAGATTGTGTGCAGGCTAGAGCTGCTCTGGATAAGGCTACCGTGCTTCTGAGTATGTCTAAAGGTGGAAAGCGTAAAGATAGTGTGTGGGGCTCTGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodong Chen et al.
Anatomical record (Hoboken, N.J. : 2007), 302(8), 1399-1408 (2018-10-20)
Osteosarcoma (OS) is one of the most common malignancies of bone. This study was aimed to explore the anti-metastatic effect of euxanthone on OS. Adhesion assay and Transwell assay were used to examine the effect of euxanthone on adhesion, migration
Xiafei Fu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 48(2), 670-682 (2018-07-20)
Several miRNAs have been reported to be involved in the pathogenesis of polycystic ovarian syndrome (PCOS). However, the biological roles of miR-16 and its molecular mechanisms in PCOS development remain to be elucidated. qRT-PCR was performed to detect the expression
Ken Watanabe et al.
PloS one, 15(2), e0226053-e0226053 (2020-02-11)
Hypertension is a major public health problem among the aging population worldwide. It causes cardiac remodeling, including hypertrophy and interstitial fibrosis, which leads to development of hypertensive heart disease (HHD). Although microRNA-21 (miR-21) is associated with fibrogenesis in multiple organs
Hongwei Liang et al.
Scientific reports, 6, 23772-23772 (2016-03-30)
Programmed cell death 4 (PDCD4), as a tumor suppressor gene, is frequently reduced in a variety of tumors, including gastric cancer. Previous findings have indicated that PDCD4 participates in tumorigenesis through the regulation of apoptosis, but the molecular basis of
Shi-Wang Wen et al.
Digestive diseases and sciences, 63(9), 2285-2293 (2018-05-22)
This study was designed to explore the anticancer potential of isoalantolactone, a sesquiterpene lactone, on esophageal squamous cell carcinoma (ESCC) cells and associated molecular mechanisms. ESCC cell lines were treated with isoalantolactone or vehicle and tested for viability, proliferation, cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service