Skip to Content
Merck
All Photos(1)

Key Documents

EHU070071

Sigma-Aldrich

MISSION® esiRNA

targeting human TNFRSF12A

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTCTGGCTTTTTGGTCTGGAGACGATGCCGCAGGAGAGAGAAGTTCACCACCCCCATAGAGGAGACCGGCGGAGAGGGCTGCCCAGCTGTGGCGCTGATCCAGTGACAATGTGCCCCCTGCCAGCCGGGGCTCGCCCACTCATCATTCATTCATCCATTCTAGAGCCAGTCTCTGCCTCCCAGACGCGGCGGGAGCCAAGCTCCTCCAACCACAAGGGGGGTGGGGGGCGGTGAATCACCTCTGAGGCCTGGGCCCAGGGTTCAGGGGAACCTTCCAAGGTGTCTGGTTGCCCTGCCTCTGGCTCCAGAACAGAAAGGGAGCCTCACGCTGGCTCACACAAAACAGCTGACACTGACTAAGGAACTGCAGCATTTGCACAGGGGAGGGGGGTGCCCTCCTTCCTAGAGGCCCTGGGGGCCAGGCTGACTTGGGGGGCAGACTTGACACTAGGCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Hao et al.
Journal of cellular and molecular medicine, 22(9), 4344-4353 (2018-07-05)
Atrial myocyte hypertrophy is one of the most important substrates in the development of atrial fibrillation (AF). The TWEAK/Fn14 axis is a positive regulator of cardiac hypertrophy in cardiomyopathy. This study therefore investigated the effects of Fn14 on atrial hypertrophy
Xuefeng Qi et al.
Frontiers in cellular and infection microbiology, 7, 315-315 (2017-07-27)
TLR4 in intestinal epithelial cells has been shown both inflammatory and homeostatic roles following binding of its cognate ligand lipopolysaccharide (LPS). TWEAK-Fn14 axis plays an important role in pathologies caused by excessive or abnormal inflammatory responses. This study aimed to
Zhengwei Li et al.
American journal of translational research, 8(12), 5386-5398 (2017-01-13)
Angiotesin II (Ang II) plays an important role in cardiac remodeling. Fibroblast growth factor inducible-14 (Fn14) is the smallest member of the tumor necrosis factor superfamily of receptors. Currently, little is known about the functional role of Fn14 in the
Xuefeng Qi et al.
The Journal of general virology, 99(1), 36-43 (2017-12-09)
The pathogenesis of H9N2 subtype avian influenza virus (AIV) infection in hens is often related to oviduct tissue damage. Our previous study suggested that H9N2 AIV induces cellular apoptosis by activating reactive oxygen species (ROS) accumulation and mitochondria-mediated apoptotic signalling
Alison Roos et al.
Molecular cancer research : MCR, 16(7), 1185-1195 (2018-05-05)
Glioblastoma multiforme (GBM) is the most common brain malignancies in adults. Most GBM patients succumb to the disease less than 1 year after diagnosis due to the highly invasive nature of the tumor, which prevents complete surgical resection and gives

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service