Skip to Content
Merck
All Photos(1)

Key Documents

EHU069501

Sigma-Aldrich

MISSION® esiRNA

targeting human SMAD2 (2)

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTCCGCCTCTGGATGACTATACTCACTCCATTCCAGAAAACACTAACTTCCCAGCAGGAATTGAGCCACAGAGTAATTATATTCCAGAAACGCCACCTCCTGGATATATCAGTGAAGATGGAGAAACAAGTGACCAACAGTTGAATCAAAGTATGGACACAGGCTCTCCAGCAGAACTATCTCCTACTACTCTTTCCCCTGTTAATCATAGCTTGGATTTACAGCCAGTTACTTACTCAGAACCTGCATTTTGGTGTTCGATAGCATATTATGAATTAAATCAGAGGGTTGGAGAAACCTTCCATGCATCACAGCCCTCACTCACTGTAGATGGCTTTACAGACCCATCAAATTCAGAGAGGTTCTGCTTAGGTTTACTCTCCAATGTTAACCGAAATGCCACGGTAGAAAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping Zhao et al.
Cancer chemotherapy and pharmacology, 84(2), 427-439 (2019-05-16)
Although DNA-mismatch-repair-deficient (dMMR) status and aberrant expression of miRNAs are both critically implicated in the pathogenesis of resistance to 5-fluorouracil (5-FU) in colorectal cancer (CRC), whether these two factors regulate tumor response to 5-FU in a coordinated manner remains unknown.
Qingde Wa et al.
Oncology reports, 39(1), 81-90 (2017-11-16)
Constitutive activation of TGF‑β signaling pathway is a well-documented mechanism responsible for the bone metastasis of prostate cancer (PCa). MicroRNAs (miRNAs) have been reported to be crucial for the activation of TGF‑β signaling via targeting downstream components of TGF‑β signaling
Wei Zhang et al.
Journal of Cancer, 12(6), 1678-1686 (2021-02-23)
Circular RNAs (circRNAs) are associated with various diseases, including cancers. However, their roles in colorectal cancer (CRC) have not been established. Hsa_circ_0000847 (circ_SMAD2) is a novel circRNA that was found to be elevated in CRC cell lines and tissues. High
Benjamin V Park et al.
Cancer discovery, 6(12), 1366-1381 (2016-09-30)
Programmed death-1 (PD-1) is a coinhibitory receptor that downregulates the activity of tumor-infiltrating lymphocytes (TIL) in cancer and of virus-specific T cells in chronic infection. The molecular mechanisms driving high PD-1 expression on TILs have not been fully investigated. We
Ayesha Ghayur et al.
American journal of physiology. Renal physiology, 317(1), F152-F162 (2019-05-30)
Glomerulonephritis (GN) is a common cause of end-stage kidney disease and is characterized by glomerular inflammation, hematuria, proteinuria, and progressive renal dysfunction. Transforming growth factor (TGF)-β is involved in glomerulosclerosis and interstitial fibrosis. TGF-β activates multiple signaling pathways, including the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service