Skip to Content
Merck
All Photos(1)

Documents

EHU068071

Sigma-Aldrich

MISSION® esiRNA

targeting human SLC8A3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACCGCTTCATGGCATCTATTGAAGTCATCACCTCTCAAGAGAGGGAGGTGACAATTAAGAAACCCAATGGAGAAACCAGCACAACCACTATTCGGGTCTGGAATGAAACTGTCTCCAACCTGACCCTTATGGCCCTGGGTTCCTCTGCTCCTGAGATACTCCTCTCTTTAATTGAGGTGTGTGGTCATGGGTTCATTGCTGGTGATCTGGGACCTTCTACCATTGTAGGGAGTGCAGCCTTCAACATGTTCATCATCATTGGCATCTGTGTCTACGTGATCCCAGACGGAGAGACTCGCAAGATCAAGCATCTACGAGTCTTCTTCATCACCGCTGCTTGGAGTATCTTTGCCTACATCTGGCTCTATATGATTCTGGCAGTCTTCTCCCCTGGTGTGGTCCAGGTTTGGGAAGGCCTCCTCACTCTCTTCTTCTTTCCAGTGTGTGTCCTTCTGGCCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giulia Di Benedetto et al.
The FEBS journal, 286(4), 737-749 (2018-12-16)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL), a cytokine belonging to the TNF superfamily, is regarded as a mediator of neurotoxicity. The constitutively expressed ion exchanger Na+ /Ca2+ exchanger isoform-3 (NCX3) has been shown to protect neurons from injury. Its expression
Guendalina Bastioli et al.
Cell death & disease, 10(2), 80-80 (2019-01-30)
Progressive accumulation of α-synuclein (α-syn) and exposure to environmental toxins are risk factors that may both concur to Parkinson's disease (PD) pathogenesis. Electrophysiological recordings of field postsynaptic potentials (fEPSPs) and Ca2+ measures in striatal brain slices and differentiated SH-SY5Y cells
Silvia Piccirillo et al.
Cell death & disease, 9(7), 731-731 (2018-06-30)
In brain ischemia, reduction in oxygen and substrates affects mitochondrial respiratory chain and aerobic metabolism, culminating in ATP production impairment, ionic imbalance, and cell death. The restoration of blood flow and reoxygenation are frequently associated with exacerbation of tissue injury

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service