Skip to Content
Merck
All Photos(1)

Key Documents

EHU063491

Sigma-Aldrich

MISSION® esiRNA

targeting human ELAVL1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGTCACCAATGTGAAAGTGATCCGCGACTTCAACACCAACAAGTGCAAAGGGTTTGGCTTTGTGACCATGACAAACTATGAAGAAGCCGCGATGGCCATAGCCAGCCTGAACGGCTACCGCCTGGGGGACAAAATCTTACAGGTTTCCTTCAAAACCAACAAGTCCCACAAATAACTCGCTCATGCTTTTTTTTGTACGGAATAGATAATTAAGAGTGAAGGAGTTGAAACTTTTCTTGTTAGTGTACAACTCATTTTGCGCCAATTTTCACAAGTGTTTGTCTTTGTCTGAATGAGAAGTGAGAAGGTTTTTATACTCTGGGATGCAACCGACATGTTCAAATGTTTGAAATCCCACAATGTTAGACCAATCTTAAGTTTCGTAAGTTATTTCCTTTAAGATATATATTAAACAGAAATCTAAGTAGAACTGCATTGACTAACCAGTCCCTCTGGATGGTGGTGAACCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prachi Matsye et al.
Glia, 65(6), 945-963 (2017-03-17)
In neurodegenerative diseases such as amyotrophic lateral sclerosis (ALS), chronic activation of microglia contributes to disease progression. Activated microglia produce cytokines, chemokines, and other factors that normally serve to clear infection or damaged tissue either directly or through the recruitment
Aya Yanagawa-Matsuda et al.
Oncology reports, 41(2), 954-960 (2018-11-16)
AU-rich elements (AREs) are RNA elements that enhance the rapid decay of mRNA. The fate of ARE-mRNA is controlled by ARE-binding proteins. HuR, a member of the embryonic lethal abnormal vision (ELAV) family of RNA-binding proteins, is involved in the
Kotb Abdelmohsen et al.
RNA biology, 14(3), 361-369 (2017-01-13)
HuR influences gene expression programs and hence cellular phenotypes by binding to hundreds of coding and noncoding linear RNAs. However, whether HuR binds to circular RNAs (circRNAs) and impacts on their function is unknown. Here, we have identified en masse
Guillaume Gauchotte et al.
The Journal of pathology, 242(4), 421-434 (2017-05-12)
HuR regulates cytoplasmic mRNA stability and translatability, and the HuR expression level has been shown to correlate with poor disease outcome in several cancer types; however, the prognostic value and potential pro-oncogenic properties of HuR in meningioma remain unclear. Thus
Jiping Wang et al.
Cancer biology & therapy, 20(7), 979-988 (2019-04-18)
Glioblastoma is a highly malignant and typically fatal tumor of the central nervous system. The tumor is characterized by marked cellular and molecular heterogeneity, including a subpopulation of brain tumor initiating cells (BTICs) that are highly resistant to radiation and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service