Skip to Content
Merck
All Photos(1)

Key Documents

EHU062341

Sigma-Aldrich

MISSION® esiRNA

targeting human C1QTNF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCATGCACAGCAACCACTACTACCAGACGGTGATCTTCGACACGGAGTTCGTGAACCTCTACGACCACTTCAACATGTTCACCGGCAAGTTCTACTGCTACGTGCCCGGCCTCTACTTCTTCAGCCTCAACGTGCACACCTGGAACCAGAAGGAGACCTACCTGCACATCATGAAGAACGAGGAGGAGGTGGTGATCTTGTTCGCGCAGGTGGGCGACCGCAGCATCATGCAAAGCCAGAGCCTGATGCTGGAGCTGCGAGAGCAGGACCAGGTGTGGGTACGCCTCTACAAGGGCGAACGTGAGAACGCCATCTTCAGCGAGGAGCTGGACACCTACATCACCTTCAGTGGCTACCTGGTCAAGCACGCCACCGAGCCCTAGCTGGCCGGCCACCTCCTTTCCTCTCGCCACCTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huilin Wang et al.
Life sciences, 256, 117992-117992 (2020-06-23)
C1q/tumor necrosis factor-related protein-1 (CTRP1) is a newly identified adiponectin paralog that modulates metabolism and inflammation. However, the cerebral function of CTRP1 remains unknown. This study aimed to determine its role and mechanism in cerebral ischemia and reperfusion injury. Serum
Leiming Wu et al.
Free radical biology & medicine, 121, 215-230 (2018-05-08)
Complement C1q tumor necrosis factor related proteins (C1QTNFs) have been reported to have diverse biological influence on the cardiovascular system. C1QTNF1 is a member of the CTRP superfamily. C1QTNF1 is expressed in the myocardium; however, its function in myocytes has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service