Skip to Content
Merck
All Photos(1)

Key Documents

EHU055781

Sigma-Aldrich

MISSION® esiRNA

targeting human LATS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTGCCTTGCTTGTAAGGTGGACACTCACGCCCTGTACGCCATGAAGACCCTAAGGAAAAAGGATGTCCTGAACCGGAATCAGGTGGCCCACGTCAAGGCCGAGAGGGACATCCTGGCCGAGGCAGACAATGAGTGGGTGGTCAAACTCTACTACTCCTTCCAAGACAAAGACAGCCTGTACTTTGTGATGGACTACATCCCTGGTGGGGACATGATGAGCCTGCTGATCCGGATGGAGGTCTTCCCTGAGCACCTGGCCCGGTTCTACATCGCAGAGCTGACTTTGGCCATTGAGAGTGTCCACAAGATGGGCTTCATCCACCGAGACATCAAGCCTGATAACATTTTGATAGATCTGGATGGTCACATTAAACTCACAGATTTCGGCCTCTGCACTGGGTTCAGGTGGACTCACAATTCCAAATATTACCAGAAAGGGAGCCATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cédric Belair et al.
Silence, 2(1), 7-7 (2011-10-27)
MicroRNAs, post-transcriptional regulators of eukaryotic gene expression, are implicated in host defense against pathogens. Viruses and bacteria have evolved strategies that suppress microRNA functions, resulting in a sustainable infection. In this work we report that Helicobacter pylori, a human stomach-colonizing
Ke Wang et al.
International journal of molecular medicine, 47(4) (2021-02-13)
Long non‑coding RNAs (lncRNAs) are a class of non‑protein coding transcripts that are involved in the regulation of gene expression in mammalian cells. Transcriptional co‑activator Yes associated protein 1 (YAP1) plays a key role in the progression of ovarian cancer.
Cong Dong et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 44(6), 475-481 (2015-03-19)
Many reports indicated LATS2 was a component of the Hippo pathway, could phosphorylate and inactivate YAP, acted as a tumor suppressor in human cancers. But few studies investigated the role of LATS2 in oral squamous cell carcinoma (OSCC) and clarified
Tone B Enger et al.
Laboratory investigation; a journal of technical methods and pathology, 93(11), 1203-1218 (2013-10-02)
Sjogren's syndrome (SS) is a complex autoimmune disease that primarily affects salivary and lacrimal glands and is associated with high morbidity. Although the prevailing dogma is that immune system pathology drives SS, increasing evidence points to structural defects, including defective
Peng Zhang et al.
Cell discovery, 2, 16021-16021 (2016-07-28)
Partitioning-defective 3 (Par3), a key component of the evolutionarily conserved polarity PAR complex (Par3/Par6/aPKC), controls cell polarity and contributes to cell migration, proliferation and tumor development. Emerging evidence indicates that cell polarity proteins function as upstream modulators that regulate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service