Skip to Content
Merck
All Photos(1)

Key Documents

EHU055711

Sigma-Aldrich

MISSION® esiRNA

targeting human RBX1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCAGGAACCACATTATGGATCTTTGCATAGAATGTCAAGCTAACCAGGCGTCCGCTACTTCAGAAGAGTGTACTGTCGCATGGGGAGTCTGTAACCATGCTTTTCACTTCCACTGCATCTCTCGCTGGCTCAAAACACGACAGGTGTGTCCATTGGACAACAGAGAGTGGGAATTCCAAAAGTATGGGCACTAGGAAAAGACTTCTTCCATCAAGCTTAATTGTTTTGTTATTCATTTAATGACTTTCCCTGCTGTTACCTAATTACAAATTGGATGGAACTGTGTTTTTTTCTGCTTTGTTTTTTCAGTTTGCTGTTTCTGTAGCCATATTGTATTCTGTGTCAAATAAAGTCCAGTTGGATTCTGGAACGGATG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiro Kunishige et al.
Oncology letters, 20(3), 2919-2927 (2020-08-13)
Ring box protein-1 (RBX1) is an essential component of the S-phase kinase-associated protein, Cullin and F-box containing ubiquitin ligases. Overexpression of RBX1 has been reported in several cancer types; however, little is known regarding the prognostic value and role of
Kazuhiro Migita et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 17(4), 601-609 (2013-12-03)
RING box protein-1 (RBX1) is an essential component of the E3 ubiquitin ligase Skp1/Cullin/RBX1/F-box protein complex. Although an altered expression of RBX1 has been reported in several human cancers, the role of RBX1 in gastric cancer remains unknown. We investigated
Fan Yao et al.
Nature communications, 9(1), 2269-2269 (2018-06-13)
Dysregulation of YAP localization and activity is associated with pathological conditions such as cancer. Although activation of the Hippo phosphorylation cascade is known to cause cytoplasmic retention and inactivation of YAP, emerging evidence suggests that YAP can be regulated in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service