Skip to Content
Merck
All Photos(1)

Key Documents

EHU051611

Sigma-Aldrich

MISSION® esiRNA

targeting human HABP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGTGGATGGTCACTGTGCTCTAGAATCCAAATACGTGAAGACTGTGTGCTTGCCTGATGGGTCCTTTCCCTCTGGGAGTGAGTGCCACATCTCTGGCTGGGGTGTTACAGAAACAGGAAAAGGGTCCCGCCAGCTCCTGGATGCCAAAGTCAAGCTGATTGCCAACACTTTGTGCAACTCCCGCCAACTCTATGACCACATGATTGATGACAGTATGATCTGTGCAGGAAATCTTCAGAAACCTGGGCAAGACACCTGCCAGGGTGACTCTGGAGGCCCCCTGACCTGTGAGAAGGACGGCACCTACTACGTCTATGGGATAGTGAGCTGGGGCCTGGAGTGTGGGAAGAGGCCAGGGGTCTACACCCAAGTTACCAAATTCCTGAATTGGATCAAAGCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamara Mirzapoiazova et al.
Frontiers in oncology, 5, 164-164 (2015-08-11)
Lung cancer is a devastating disease with limited treatment options. Many lung cancers have changes in their microenvironment including upregulation of the extracellular matrix glycosaminoglycan, hyaluronan (HA), which we have previously demonstrated can regulate the activity of the extracellular serine
Sudheer Kumar Gara et al.
The New England journal of medicine, 373(5), 448-455 (2015-07-30)
Familial nonmedullary thyroid cancer accounts for 3 to 9% of all cases of thyroid cancer, but the susceptibility genes are not known. Here, we report a germline variant of HABP2 in seven affected members of a kindred with familial nonmedullary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service