Skip to Content
Merck
All Photos(1)

Documents

EHU048531

Sigma-Aldrich

MISSION® esiRNA

targeting human VAV1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGCCACATGTTCCTCCTGATCGAGGACCAAGGTGCCCAGGGCTATGAGCTGTTCTTCAAGACAAGAGAATTGAAGAAGAAGTGGATGGAGCAGTTTGAGATGGCCATCTCCAACATCTATCCGGAGAATGCCACCGCCAACGGGCATGACTTCCAGATGTTCTCCTTTGAGGAGACCACATCCTGCAAGGCCTGTCAGATGCTGCTTAGAGGTACCTTCTATCAGGGCTACCGCTGCCATCGGTGCCGGGCATCTGCACACAAGGAGTGTCTGGGGAGGGTCCCTCCATGTGGCCGACATGGGCAAGATTTCCCAGGAACTATGAAGAAGGACAAACTACATCGCAGGGCTCAGGACAAAAAGAGGAATGAGCTGGGTCTGCCCAAGATGGAGGTGTTTCAGGAATACTACGGGCTTCCTCCACCCCCTGGAGCCATTGGACCCTTTCTACGGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Zhou et al.
Journal of cellular biochemistry, 119(7), 5437-5448 (2018-01-26)
This study aims to explore the effect of miR-330 targeting VAV1 on amyloid β-protein (Aβ) production, oxidative stress (OS), and mitochondrial dysfunction in Alzheimer's disease (AD) mice through the MAPK signaling pathway. Putative targeted gene of miR-330 was performed by
Guillaume Gaud et al.
Science signaling, 11(538) (2018-07-12)
The activation of T cells requires the guanine nucleotide exchange factor VAV1. Using mice in which a tag for affinity purification was attached to endogenous VAV1 molecules, we analyzed by quantitative mass spectrometry the signaling complex that assembles around activated
Arathi Nair et al.
Cell communication and signaling : CCS, 18(1), 3-3 (2020-01-08)
Ras are small cellular GTPases which regulate diverse cellular processes. It has three isoforms: H-Ras, K-Ras, and N-Ras. Owing to the N-terminus (1-165 residues) sequence homology these isoforms were thought to be functionally redundant. However, only K-Ras-deficient mice but not
Silvia Grassilli et al.
Oncotarget, 5(12), 4320-4336 (2014-06-26)
Vav1 is one of the signalling proteins normally restricted to hematopoietic cells that results ectopically expressed in solid tumors, including breast cancer. By immunohistochemical analysis on TMAs containing invasive breast tumor from patients without lymph node involvement, we have found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service