Skip to Content
Merck
All Photos(1)

Key Documents

EHU042391

Sigma-Aldrich

MISSION® esiRNA

targeting human ETHE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGTGTCACCTTCGTCCTGAATGACCACAGCATGGCCTTCACTGGAGATGCCCTGTTGATCCGTGGGTGTGGGCGGACAGACTTCCAGCAAGGCTGTGCCAAGACCTTGTACCACTCGGTCCATGAAAAGATCTTCACACTTCCAGGAGACTGTCTGATCTACCCTGCTCACGATTACCATGGGTTCACAGTGTCCACCGTGGAGGAGGAGAGGACTCTGAACCCTCGGCTCACCCTCAGCTGTGAGGAGTTTGTCAAAATCATGGGCAACCTGAACTTGCCTAAACCTCAGCAGATAGACTTTGCTGTTCCAGCCAACATGCGCTGTGGGGTGCAGACACCCACTGCCTGATCTCACTTCTGTCAGATGCTCCCATCCACTATTAATGCACTAGGTGGGAGGAGAGGGCGGCAATGACACTGCACCTCTCCTTTCCCACCGCATTCCCTGGAGCTCCCTAAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuhui Hao et al.
Scientific reports, 6, 38942-38942 (2016-12-15)
The purpose of this study was to investigate the underlying mechanism of metallothionein (MT) protection from depleted uranium (DU) using a proteomics approach to search for a DU toxicity-differential protein. MT-/- and MT+/+ mice were administrated with a single dose
Yuhui Hao et al.
Journal of biochemical and molecular toxicology, 35(3), e22669-e22669 (2020-12-05)
The kidney is the target of the acute toxicity of depleted uranium (DU). However, the mechanism of DU-induced cytotoxicity is not clear. The study was to demonstrate the role of autophagy in DU-induced cytotoxicity and to determine the potential mechanism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service