Skip to Content
Merck
All Photos(1)

Documents

EHU040491

Sigma-Aldrich

MISSION® esiRNA

targeting human BMP6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GATGGCAGGACTGGATCATTGCACCCAAGGGCTATGCTGCCAATTACTGTGATGGAGAATGCTCCTTCCCACTCAACGCACACATGAATGCAACCAACCACGCGATTGTGCAGACCTTGGTTCACCTTATGAACCCCGAGTATGTCCCCAAACCGTGCTGTGCGCCAACTAAGCTAAATGCCATCTCGGTTCTTTACTTTGATGACAACTCCAATGTCATTCTGAAAAAATACAGGAATATGGTTGTAAGAGCTTGTGGATGCCACTAACTCGAAACCAGATGCTGGGGACACACATTCTGCCTTGGATTCCTAGATTACATCTGCCTTAAAAAAACACGGAAGCACAGTTGGAGGTGGGACGATGAGACTTTGAAACTATCTCATGCCAGTGCCTTATTACCCAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica C Rivera et al.
Frontiers in pharmacology, 4, 125-125 (2013-10-09)
Bone morphogenetic proteins (BMPs) other than the clinically available BMP-2 and BMP-7 may be useful for improving fracture healing through both increasing osteogenesis and creating a favorable healing environment by altering cytokine release by endogenous cells. Given the spectrum of
Geng Liu et al.
Oncology reports, 32(2), 581-588 (2014-06-04)
Bone morphogenetic protein 6 (BMP6) is an important regulator of cell growth, differentiation and apoptosis in various types of tumor. In breast cancer, it was considered as a tumor suppressor. Our previous study also confirmed that BMP6 was a critical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service