Skip to Content
Merck
All Photos(1)

Key Documents

EHU020001

Sigma-Aldrich

MISSION® esiRNA

targeting human IRF3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€197.00
50 μG
€349.00

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
20 μG
€197.00
50 μG
€349.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€197.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GATGCACAGCAGGAGGATTTCGGAATCTTCCAGGCCTGGGCCGAGGCCACTGGTGCATATGTTCCCGGGAGGGATAAGCCAGACCTGCCAACCTGGAAGAGGAATTTCCGCTCTGCCCTCAACCGCAAAGAAGGGTTGCGTTTAGCAGAGGACCGGAGCAAGGACCCTCACGACCCACATAAAATCTACGAGTTTGTGAACTCAGGAGTTGGGGACTTTTCCCAGCCAGACACCTCTCCGGACACCAATGGTGGAGGCAGTACTTCTGATACCCAGGAAGACATTCTGGATGAGTTACTGGGTAACATGGTGTTGGCCCCACTCCCAGATCCGGGACCCCCAAGCCTGGCTGTAGCCCCTGAGCCCTGCCCTCAGCCCCTGCGGAGCCCCAGCTTGGACAATCCCACTCCCTTCCCAAACCTGGGGCCCTCTGAGAACCCACTGAAGCGGCTGTTGGTGCCGGGGGAAGAGTGGGAGTTCGAGGTGACAGCCTTCTACCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanae Harashima et al.
Molecular cancer, 13, 217-217 (2014-09-18)
Synthetic double-stranded RNA poly(I:C) is a useful immune adjuvant and exhibits direct antitumor effects against several types of cancers. In this study, we elucidated the mechanisms underlying the effects induced in poly(I:C)-transfected human renal cell carcinoma (RCC) cells. In contrast
Xiao-Hua Wang et al.
The international journal of biochemistry & cell biology, 87, 8-17 (2017-03-25)
Respiratory syncytial virus (RSV) is the leading cause of bronchiolitis in infancy, which is a major risk factor for recurrent wheezing and asthma. Orosomucoid 1-like protein 3 (ORMDL3) has been reported to associate with virus-triggered recurrent wheezing and asthma in
Iris F Ueki et al.
The Journal of experimental medicine, 210(10), 1929-1936 (2013-09-04)
Viruses suppress host responses to increase infection, and understanding these mechanisms has provided insights into cellular signaling and led to novel therapies. Many viruses (e.g., Influenza virus, Rhinovirus [RV], Cytomegalovirus, Epstein-Barr virus, and Hepatitis C virus) activate epithelial epidermal growth
Ning Li et al.
Redox biology, 24, 101215-101215 (2019-05-24)
Mountainous evidence suggests that inflammation, cardiomyocyte apoptosis and pyroptosis are involved in the development of sepsis and sepsis-induced cardiomyopathy (SIC). Stimulator of interferon genes (STING) is an indispensable molecule that could regulate inflammation and immune response in multiple diseases. However
Koji Hirono et al.
Modern rheumatology, 30(6), 1074-1081 (2019-10-19)
Background: Endothelial expression of membrane-bound fractalkine/CX3CL1 (Fkn) reportedly acts as a strong mediator of inflammation. Toll-like receptor 3 (TLR3) axes are thought to play some roles in the development of chronic glomerulonephritis (CGN) including lupus nephritis (LN). However, detailed mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service