Skip to Content
Merck
All Photos(1)

Key Documents

EHU016751

Sigma-Aldrich

MISSION® esiRNA

targeting human CTGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGCACCAGCATGAAGACATACCGAGCTAAATTCTGTGGAGTATGTACCGACGGCCGATGCTGCACCCCCCACAGAACCACCACCCTGCCGGTGGAGTTCAAGTGCCCTGACGGCGAGGTCATGAAGAAGAACATGATGTTCATCAAGACCTGTGCCTGCCATTACAACTGTCCCGGAGACAATGACATCTTTGAATCGCTGTACTACAGGAAGATGTACGGAGACATGGCATGAAGCCAGAGAGTGAGAGACATTAACTCATTAGACTGGAACTTGAACTGATTCACATCTCATTTTTCCGTAAAAATGATTTCAGTAGCACAAGTTATTTAAATCTGTTTTTCTAACTGGGGGAAAAGATTCCCACCCAATTCAAAACATTGTGCCATGTCAAACAAATAGTCTATCAACCCCAGACACTGGTTTGAAGAATGTTAAGACTTGACAGTGGAACTACATTAGTACACAGCACCAGAATGTATATTAAGGTGTGGCTTTAGGAGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rohan Samarakoon et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(10), 3308-3320 (2016-06-23)
Protein phosphatase magnesium-dependent-1A (PPM1A) dephosphorylates SMAD2/3, which suppresses TGF-β signaling in keratinocytes and during Xenopus development; however, potential involvement of PPM1A in chronic kidney disease is unknown. PPM1A expression was dramatically decreased in the tubulointerstitium in obstructive and aristolochic acid
Jung-Chien Cheng et al.
Oncotarget, 8(49), 85224-85233 (2017-11-22)
Ovarian low-grade serous carcinoma (LGSC) is a rare disease and is now considered to be a distinct entity from high-grade serous carcinoma (HGSC), which is the most common and malignant form of epithelial ovarian cancer. Connective tissue growth factor (CTGF)
Hiroshi Kinashi et al.
Scientific reports, 9(1), 12175-12175 (2019-08-23)
Lymphatic absorption in the peritoneal cavity may contribute to ultrafiltration failure in peritoneal dialysis (PD). Lymphatic vessels develop during PD-related peritoneal fibrosis. Connective tissue growth factor (CTGF, also called CCN2) is an important determinant of fibrotic tissue remodeling, but little
Kai Yang et al.
OncoTargets and therapy, 9, 7285-7295 (2016-12-13)
Colorectal cancer (CRC) is one of the most commonly diagnosed cancers among both males and females; the chemotherapy drug 5-fluorouracil (5-FU) is one of a doctors' first lines of defense against CRC. However, therapeutic failures are common because of the
Huijun Zeng et al.
Cell death & disease, 8(6), e2885-e2885 (2017-06-16)
Limited benefits and clinical utility of temozolomide (TMZ) for glioblastoma (GB) are frequently compromised by the development of acquired drug resistance. Overcoming TMZ resistance and uncovering the underlying mechanisms are challenges faced during GB chemotherapy. In this study, we reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service