Skip to Content
Merck
All Photos(1)

Key Documents

EHU010941

Sigma-Aldrich

MISSION® esiRNA

targeting human CYBB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTTGTGGCTGTGATAAGCAGGAGTTTCAAGATGCGTGGAAACTACCTAAGATAGCGGTTGATGGGCCCTTTGGCACTGCCAGTGAAGATGTGTTCAGCTATGAGGTGGTGATGTTAGTGGGAGCAGGGATTGGGGTCACACCCTTCGCATCCATTCTCAAGTCAGTCTGGTACAAATATTGCAATAACGCCACCAATCTGAAGCTCAAAAAGATCTACTTCTACTGGCTGTGCCGGGACACACATGCCTTTGAGTGGTTTGCAGATCTGCTGCAACTGCTGGAGAGCCAGATGCAGGAAAGGAACAATGCCGGCTTCCTCAGCTACAACATCTACCTCACTGGCTGGGATGAGTCTCAGGCCAATCACTTTGCTGTGCACCATGATGAGGAGAAAGATGTGATCACAGGCCTGAAACAAAAGACTTTGTATGGACGGCCCAACTGGGATAATGAATTCAAGACAATTGCAAGTCAACACCCTAATACCAGAATAGGAGTTTTCCTCTGTGGACCTGAAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjun Wang et al.
Brain research bulletin, 160, 141-149 (2020-05-12)
Sleep deprivation (SD) can induce cognitive and memory impairments. This impairment is in part due to oxidative stress damage in the hippocampus region of the brain. Corilagin (CL), a polyphenol belonging to the tannin family and extracted from Terminalia chebula
Young-Mee Kim et al.
American journal of physiology. Cell physiology, 312(6), C749-C764 (2017-04-21)
Reactive oxygen species (ROS) derived from NADPH oxidase (NOX) and mitochondria play a critical role in growth factor-induced switch from a quiescent to an angiogenic phenotype in endothelial cells (ECs). However, how highly diffusible ROS produced from different sources can
Xianzhang Zeng et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 50(2), 783-797 (2018-10-15)
Peri-operative cerebral ischemia reperfusion injury is one of the most serious peri-operative complications that can be aggravated in patients with diabetes. A previous study showed that microglia NOX2 (a NADPH oxidase enzyme) may play an important role in this process.
Yongpan Huang et al.
Oxidative medicine and cellular longevity, 2020, 3912173-3912173 (2020-12-05)
Oxymatrine (OMT) is the major quinolizidine alkaloid extracted from the root of Sophora flavescens Ait and has been shown to exhibit a diverse range of pharmacological properties. The aim of the present study was to investigate the role of OMT
Christopher R Marlein et al.
Blood, 130(14), 1649-1660 (2017-07-25)
Improvements in the understanding of the metabolic cross-talk between cancer and its microenvironment are expected to lead to novel therapeutic approaches. Acute myeloid leukemia (AML) cells have increased mitochondria compared with nonmalignant CD34

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service