Skip to Content
Merck
All Photos(1)

Documents

EHU008751

Sigma-Aldrich

MISSION® esiRNA

targeting human EPAS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGGCCAGGTGAAAGTCTACAACAACTGCCCTCCTCACAATAGTCTGTGTGGCTACAAGGAGCCCCTGCTGTCCTGCCTCATCATCATGTGTGAACCAATCCAGCACCCATCCCACATGGACATCCCCCTGGATAGCAAGACCTTCCTGAGCCGCCACAGCATGGACATGAAGTTCACCTACTGTGATGACAGAATCACAGAACTGATTGGTTACCACCCTGAGGAGCTGCTTGGCCGCTCAGCCTATGAATTCTACCATGCGCTAGACTCCGAGAACATGACCAAGAGTCACCAGAACTTGTGCACCAAGGGTCAGGTAGTAAGTGGCCAGTACCGGATGCTCGCAAAGCATGGGGGCTACGTGTGGCTGGAGACCCAGGGGACGGTCATCTACAACCCTCGCAACCTGCAGCCCCAGTGCATCATGTGTGTCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seungyeul Yoo et al.
PLoS genetics, 11(1), e1004898-e1004898 (2015-01-09)
Chronic Obstructive Pulmonary Disease (COPD) is a complex disease. Genetic, epigenetic, and environmental factors are known to contribute to COPD risk and disease progression. Therefore we developed a systematic approach to identify key regulators of COPD that integrates genome-wide DNA
Kei Houri et al.
Scientific reports, 10(1), 3735-3735 (2020-03-01)
Elevation of the levels of reactive oxygen species (ROS) is a major tissue-degenerative phenomenon involved in aging and aging-related diseases. The detailed mechanisms underlying aging-related ROS generation remain unclear. Presently, the expression of microRNA (miR)-142-5p was significantly upregulated in bone
Olga Roche et al.
Nucleic acids research, 44(19), 9315-9330 (2016-11-02)
A wide range of diseases course with an unbalance between the consumption of oxygen by tissues and its supply. This situation triggers a transcriptional response, mediated by the hypoxia inducible factors (HIFs), that aims to restore oxygen homeostasis. Little is
Farhadul Islam et al.
Frontiers in oncology, 10, 1534-1534 (2020-10-13)
Endothelial PAS domain-containing protein 1 (EPAS1) is an angiogenic factor and its implications have been reported in many cancers but not in esophageal squamous cell carcinoma (ESCC). Herein, we aim to examine the genetic and molecular alterations, clinical implications, and
Shirley Dehn et al.
Journal of immunology (Baltimore, Md. : 1950), 197(9), 3639-3649 (2016-09-28)
Hypoxia-inducible factor (HIF)-α isoforms regulate key macrophage (MΦ) functions during ischemic inflammation. HIF-2α drives proinflammatory cytokine production; however, the requirements for HIF-2α during other key MΦ functions, including phagocytosis, are unknown. In contrast to HIF-1α, HIF-2α was not required for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service